Skip to Content
Merck
All Photos(2)

Key Documents

D7750

Sigma-Aldrich

2′-Deoxycytidine 5′-monophosphate

Sigma Grade, ≥95.0%

Synonym(s):

Deoxycytidylic acid, dCMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N3O7P
CAS Number:
Molecular Weight:
307.20
Beilstein:
41062
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic

grade

Sigma Grade

Assay

≥95.0%

form

powder

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

NC1=NC(=O)N(C=C1)[C@H]2C[C@H](O)[C@@H](COP(O)(O)=O)O2

InChI

1S/C9H14N3O7P/c10-7-1-2-12(9(14)11-7)8-3-5(13)6(19-8)4-18-20(15,16)17/h1-2,5-6,8,13H,3-4H2,(H2,10,11,14)(H2,15,16,17)/t5-,6+,8+/m0/s1

InChI key

NCMVOABPESMRCP-SHYZEUOFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′-Deoxycytidine 5′-monophosphate is a deoxynucleotide building block via which DNA is made. It consists of a 2′-deoxy-β-D-ribofuranose, linked through an N-β-D-glycosidic linkage to a heterocycle nitrogen cytosine. It is phosphorylated at carbon C-5′.

Application

2′-Deoxycytidine 5′-monophosphate (dCMP) is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA biosynthesis.
2′-Deoxycytidine 5′-monophosphate has been used in Raman spectroscopic studies to determine its Raman total half bandwidths.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivan Zlatev et al.
Bioorganic & medicinal chemistry, 17(19), 7008-7014 (2009-09-01)
The replacement of the pyrophosphate moiety of 2'-deoxynucleoside triphosphates by non natural delta-dicarboxylic butyl amino acid allows incorporation of natural 2'-deoxycytidine into DNA using HIV-1 reverse transcriptase (RT) as enzyme. In contrast, the 3'-deoxycytidine analogue was not a substrate of
Susan Quinn et al.
Chemical communications (Cambridge, England), (21)(21), 2130-2132 (2007-05-24)
A strong infrared band at 1574 cm(-1) is observed following 267 nm excitation of 2'-deoxycytidine (tau = 37 +/- 4 ps) or 2'-deoxycytidine 5'-monophosphate (tau = 33 +/- 4 ps); this band is provisionally attributed to an 1n(N)pi* state and
Makusu Tsutsui et al.
Journal of the American Chemical Society, 133(23), 9124-9128 (2011-05-13)
We report label-free electrical detections of chemically modified nucleobases in a DNA using a nucleotide-sized electrode gap. We found that methyl substitution contributes to increase the tunneling conductance of deoxycytidines, which was attributed to a shift of the highest occupied
Montserrat Terrazas et al.
Chemistry & biodiversity, 5(1), 31-39 (2008-01-22)
The replacement of the pyrophosphate moiety of deoxynucleoside triphosphates by L-aspartic acid allows incorporation of natural deoxynucleosides into DNA using HIV reverse transcriptase (RT) as enzyme, while retaining the canonical base-pair selectivity. N-Methylation of the L-aspartic acid leaving group results
Zimin Liu et al.
American journal of translational research, 8(11), 4764-4777 (2016-12-03)
microRNAs (miRs) are small noncoding RNAs that modulate a variety of cellular processes by regulating multiple targets, which can promote or inhibit the development of malignant behaviors. Accumulating evidence suggests that microRNA-221 (miR-221) plays important roles in human carcinogenesis. It

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service