D7321
Diosmetin
Synonym(s):
3′,5,7-Trihydroxy-4′-methoxyflavone, 4′-Methylluteolin, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chromen-4-one, Luteolin 4′-methyl ether
About This Item
Recommended Products
Assay
≥98% (HPLC)
Quality Level
form
powder
color
light yellow to yellow
solubility
DMSO: 1 mg/mL, clear, faintly yellow to greenish-yellow
application(s)
metabolomics
vitamins, nutraceuticals, and natural products
storage temp.
2-8°C
SMILES string
COc1ccc(cc1O)C2=CC(=O)c3c(O)cc(O)cc3O2
InChI
1S/C16H12O6/c1-21-13-3-2-8(4-10(13)18)14-7-12(20)16-11(19)5-9(17)6-15(16)22-14/h2-7,17-19H,1H3
InChI key
MBNGWHIJMBWFHU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service