Skip to Content
Merck
All Photos(1)

Key Documents

D7291

Sigma-Aldrich

Deoxyribonuclease I RNase-free solution from bovine pancreas

Synonym(s):

DNase I, Deoxyribonuclease I, pancreatic DNase I, pancreatic deoxyribonuclease I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.53

biological source

bovine pancreas

grade

Molecular Biology
for molecular biology

form

buffered aqueous glycerol solution

specific activity

10000 units/mg protein





mol wt

29.1 kDa

concentration

5—15 mg protein/mL

technique(s)

DNA extraction: suitable

suitability

suitable for molecular biology

UniProt accession no.

application(s)

cell analysis

foreign activity

RNase, none detected

storage temp.

−20°C

Gene Information

General description

Deoxyribonuclease I digests single- and double-stranded DNA to a mixture of mono and oligonucleotides carrying 5′phosphates and 3′OH termini. This catalytic activity is divalent ion-dependent. In the presence of Mg2+, DNase I hydrolyzes each strand of double-stranded DNA randomly and independently. In the presence of Mn2+, both strands can be cleaved.

Application

Deoxyribonuclease I RNase-free solution from bovine pancreas has been used to digest DNA from various samples.
Used in molecular biology applications for removing DNA during RNA purification, for preparing DNA for nick translation, and for DNA-protein interaction analysis by footprinting methods.

Components

DNase I is provided in a solution of 50% (v/v) glycerol, 20 mM sodium acetate (pH 6.5), 5 mM CaCl2, and 0.1 mM PMSF.

Unit Definition

One unit will cause a ΔA260 of 0.001 per min per mL reaction mixture using calf thymus DNA as substrate.

Other Notes

RNA treated with this DNase I should not be used to generate a cDNA library or used for RT-PCR. For these sensitive applications, it is recommended to use DNase I, Amplification Grade , Catalog Number AMP-D1.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Homburg et al.
The Journal of cell biology, 150(2), 293-307 (2000-07-26)
We present the first evidence for a fast activation of the nuclear protein poly(ADP-ribose) polymerase (PARP) by signals evoked in the cell membrane, constituting a novel mode of signaling to the cell nucleus. PARP, an abundant, highly conserved, chromatin-bound protein
Expression of anti-Mullerian hormone and its type II receptor in germ cells of maturing rat testis
Ohyama K, et al.
Endocrine Journal, EJ15-0370 (2015)
Umesh Kumar Dhawan et al.
Arteriosclerosis, thrombosis, and vascular biology, 41(10), 2598-2615 (2021-08-06)
Objective: Hypercholesterolemia-induced NETosis and accumulation of neutrophil extracellular traps (NETs) in the atherosclerotic lesion exacerbates inflammation and is causally implicated in plaque progression. We investigated whether hypercholesterolemia additionally impairs the clearance of NETs mediated by endonucleases such as DNase1 and
Stage-Specific Expression Profiles of Anti-Mullerian Hormone and its Type II Receptor in Germ Cells during Spermatogenic Cycle of Rats
Ohyama K, et al.
Journal of Steroids & Hormonal Science, 8(187), 2-2 (2017)
A Deoxyribonuclease from Chlamydomonas reinhardii:Purification and Properties
TAIT GCL and HARRIS WJ
European Journal of Biochemistry, 75(2), 357-364 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service