Skip to Content
Merck
All Photos(2)

Key Documents

D5319

Sigma-Aldrich

Deoxyribonuclease I bovine

recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein

Synonym(s):

DNAse I, Deoxyribonucleate 5′-oligonucleotido-hydrolase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bovine

Quality Level

recombinant

expressed in Pichia pastoris

Assay

≥95%

form

buffered aqueous glycerol solution

specific activity

≥5,000 units/mg protein

mol wt

~39 kDa

technique(s)

DNA extraction: suitable

suitability

suitable for molecular biology

application(s)

diagnostic assay manufacturing

foreign activity

RNAse and protease, free

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

DNAse I from Sigma was used to treat nuclear lysate to obtain single nucleosomes in a study.[1] The enzyme has also been for the preparation and harvest of mice mammary glands.[2]
Deoxyribonuclease I bovine has been used in a study to compare the initial actions of spleen deoxyribonuclease and pancreatic deoxyribonuclease. Deoxyribonuclease I bovine has also been used in a study to investigate deoxythymidine 3′, 5′-di-p-nitrophenyl phosphate as a synthetic substrate for bovine pancreatic deoxyribonuclease.
Used for the removal of DNA from protein samples.

Biochem/physiol Actions

DNase I is an endonuclease that acts on phosphodiester bonds (adjacent to pyrimidines) to produce polynucleotides with terminal 5′-phosphates. The pH optimum is found to be between 7 and 8. Divalent cations such as Mn2+, Ca2+, Co2+, and Zn2+ are activators of the enzyme. A concentration of 5 mM Ca2+ stabilizes the enzyme against proteolytic digestion.[3] 2-Mercaptoethanol, chelators, sodium dodecyl sulfate (SDS)[4] and actin[5] are known to inhibit the enzyme activity.
Digests single- and double-stranded DNA to a mixture of mono- and oligonucleotides carrying 5′ phosphates and 3′ OH termini. This catalytic activity is divalent ion-dependent. In the presence of Mg2+, DNase I hydrolyzes each strand of double-stranded DNA randomly and independently. In the presence of Mn2+, both strands can be cleaved.

Features and Benefits

  • RNA purification by removing DNA
  • Prepare DNA for nick translation1
  • Footprinting assays to determine DNA-protein interactions2

Unit Definition

One unit will produce a ΔA260 of 0.001 per min per mL reaction mixture using calf thymus DNA at pH 5.0 and 25°C

Physical form

Supplied as a solution in 4 mg/ml glycine pH 5.0, 5 mM calcium acetate and 50% glycerol

Preparation Note

Produced without using any animal cells or animal derived materials.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wendy A Kellner et al.
Nucleic acids research, 41(20), 9274-9283 (2013-08-16)
Brd4 is a double bromodomain protein that has been shown to interact with acetylated histones to regulate transcription by recruiting Positive Transcription Elongation Factor b to the promoter region. Brd4 is also involved in gene bookmarking during mitosis and is
Hongyuan Xing et al.
Journal of nanobiotechnology, 19(1), 264-264 (2021-09-08)
Exosome therapy is a promising therapeutic approach for intervertebral disc degeneration (IVDD) and achieves its therapeutic effects by regulating metabolic disorders, the microenvironment and cell homeostasis with the sustained release of microRNAs, proteins, and transcription factors. However, the rapid clearance
Alice S Kaanta et al.
Breast cancer research : BCR, 15(4), R65-R65 (2013-08-21)
The mouse mammary gland provides a powerful model system for studying processes involved in epithelial tissue development. Although markers that enrich for mammary stem cells and progenitors have been identified, our understanding of the mammary developmental hierarchy remains incomplete. We
Properties of chromatographically purified bovine pancreatic deoxyribonuclease.
P A Price et al.
The Journal of biological chemistry, 244(3), 917-923 (1969-02-10)
Heidi Hempel Sullivan et al.
The Journal of pathology, 253(4), 415-426 (2020-12-19)
We reported previously that high numbers of mast cells in benign (extra-tumoral) regions of the prostate are associated with worse outcomes after radical prostatectomy including biochemical recurrence and the development of metastases. Herein, with a cohort of 384 men, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service