Skip to Content

D3196

Dipeptidyl Peptidase VIII human

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.54
UNSPSC Code:
12352204
Specific activity:
≥20 units/μg protein
Recombinant:
expressed in Sf9 cells
Concentration:
≥0.08 mg/mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

recombinant

expressed in Sf9 cells

form

solution

specific activity

≥20 units/μg protein

mol wt

100 kDa

concentration

≥0.08 mg/mL

shipped in

dry ice

storage temp.

−70°C

General description

Full length sequence with an N-terminal GST tag

Application

Human dipeptidyl peptidase VIII has been used in a study to identify ERGIC-53 as an intracellular transport receptor of α1-antitrypsin. Human dipeptidyl peptidase VIII has also been used in a study to investigate the cargo selectivity of the ERGIC-53/MCFD2 transport receptor complex.

Biochem/physiol Actions

Dipeptidyl peptidase VIII (DPP8) is a 100 kDa monomeric protein expressed in the cytoplasm. It is a postproline dipeptidyl aminopeptidase that is homologous to DPPIV and fibroblast activation protein (FAP). It hydrolyzes Ala-Pro, Arg-Pro and Gly-Pro. DPP8 has a neutral pH optimum. It may be involved in T-cell activation and immune function as it is similar to DPPIV . It is upregulated during immune activation.

Physical form

Supplied as a solution in 40 mM Tris-HCL, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 3 mM DTT and 20% glycerol.

Other Notes

One unit will hydrolyze 1.0 picomole of Ala-Pro-AMC per minute at pH 7.4 at 25 deg °C


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library