Skip to Content
Merck
All Photos(1)

Key Documents

D1821

Sigma-Aldrich

DAF-FM

≥98% (HPLC)

Synonym(s):

4-Amino-5-methylamino-2′,7′-difluorescein, Diaminofluorescein-FM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H14F2N2O5
CAS Number:
Molecular Weight:
412.34
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to brown

solubility

DMSO: >1 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

CNc1ccc2c(c1N)C(=O)OC23c4cc(F)c(O)cc4Oc5cc(O)c(F)cc35

InChI

1S/C21H14F2N2O5/c1-25-13-3-2-8-18(19(13)24)20(28)30-21(8)9-4-11(22)14(26)6-16(9)29-17-7-15(27)12(23)5-10(17)21/h2-7,25-27H,24H2,1H3

InChI key

DIJCILWNOLHJCG-UHFFFAOYSA-N

Application

DAF-FM has been used in the visualization of nitric oxide (NO) in root sections of wheat seedlings. It has also been used to label neural crest (NC) cells.

Biochem/physiol Actions

DAF-FM is a highly sensitive, photo-stable fluorescent probe for the detection of nitric oxide (NO).
DAF-FM is a highly sensitive, photo-stable fluorescent probe for the detection of nitric oxide (NO). DAF-2 is the original compound and is widely used, but DAF-FM is more sensitive to nitric oxide (NO) compared with DAF-2. DAF-FM is more photo-stable and less pH sensitive than DAF-2. DAF-FM is not quite as cell-permeable as the diacetate derivative, DAF-FM, but it does not need to be activated by cytosolic enzymes.

Caution

hygroscopic

Preparation Note

Color variation within the range from white to brown does not change performance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visualization of Nitric Oxide, Measurement of Nitrosothiols Content, Activity of NOS and NR in Wheat Seedlings
Adavi SB, et al.
Bio-protocol, 9(20) (2019)
Phillip Roder et al.
Scientific reports, 8(1), 5892-5892 (2018-04-14)
Local manipulation of complex tissues at the single-cell level is challenging and requires excellent sealing between the specimen and the micromanipulation device. Here, biological applications for a recently developed loading technique for a force- and pressure-controlled fluidic force microscope micropipette
Kunal Bhattacharya et al.
Advanced healthcare materials, 7(6), e1700916-e1700916 (2018-01-16)
Polyethylene glycol (PEG)-modified carbon nanotubes have been successfully employed for intra-articular delivery in mice without systemic or local toxicity. However, the fate of the delivery system itself remains to be understood. In this study 2 kDa PEG-modified single-walled carbon nanotubes
Judy Choi et al.
Environmental health perspectives, 118(5), 589-595 (2010-05-05)
Silica nanoparticles (SiNPs) are being formulated for cellular imaging and for nonviral gene delivery in the central nervous system (CNS), but it is unclear what potential effects SiNPs can elicit once they enter the CNS. As the resident macrophages of
Fernando Toshio Ogata et al.
PloS one, 8(12), e84588-e84588 (2014-01-01)
Thioredoxin (TRX-1) is a multifunctional protein that controls the redox status of other proteins. TRX-1 can be found in the extracellular milieu, cytoplasm and nucleus, and it has distinct functions in each environment. Previously, we studied the intracellular localization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service