Skip to Content
Merck
All Photos(1)

Key Documents

D1446

Sigma-Aldrich

Dexrazoxane

≥95% (HPLC)

Synonym(s):

(+)-(S)-4,4′-Propylenedi-2,6-piperazinedione, (S)-(+)-1,2-Bis(3,5-dioxopiperazin-1-yl)propane, Cardioxane, ICRF-187, NSC169780, Zinecard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N4O4
CAS Number:
Molecular Weight:
268.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

originator

Johnson & Johnson

storage temp.

room temp

SMILES string

C[C@@H](CN1CC(=O)NC(=O)C1)N2CC(=O)NC(=O)C2

InChI

1S/C11H16N4O4/c1-7(15-5-10(18)13-11(19)6-15)2-14-3-8(16)12-9(17)4-14/h7H,2-6H2,1H3,(H,12,16,17)(H,13,18,19)/t7-/m0/s1

InChI key

BMKDZUISNHGIBY-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dexrazoxane is a member of bis(2,6-dioxopiperazines), that functions as a topoisomerase 2 catalytic inhibitor. Dexrazoxane is a free radical scavenger. It might protect the heart from doxorubicin-associated damage. Dexrazoxane acts as a cardiopulmonary protectant, while treating Hodgkin′s disease (HD). It functions as a chelating agent, which limits the formation of anthracycline-iron complexes. It is used to synthesize antimalarial drugs.

Application

Dexrazoxane has been used in chromatin remodelling experiments.

Biochem/physiol Actions

Dexrazoxane is a cardioprotective compound against anthracyclines. It functions by inhibiting topoisomerase II without inducing DNA strand breaks. Dexrazoxane is a + enantiomer of razoxane.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Topoisomerase IIβ-mediated DNA double-strand breaks: implications in doxorubicin cardiotoxicity and prevention by dexrazoxane
Lyu YL, et al.
Cancer Research, 67(18), 8839-8846 (2007)
The effect of dexrazoxane on myocardial injury in doxorubicin-treated children with acute lymphoblastic leukemia
Lipshultz SE, et al.
The New England Journal of Medicine, 351(2), 145-153 (2004)
Dexrazoxane-associated risk for acute myeloid leukemia/myelodysplastic syndrome and other secondary malignancies in pediatric Hodgkin's disease
Tebbi CK, et al.
Journal of Clinical Oncology, 25(5), 493-500 (2007)
Volume Transitions of Isolated Cell Nuclei Induced by Rapid Temperature Increase
Chan CJ, et al.
Biophysical Journal, 112(6), 1063-1076 (2017)
Plasmodium falciparum and Plasmodium yoelii: Effect of the Iron Chelation Prodrug Dexrazoxane onin VitroCultures
Loyevsky M, et al.
Experimental Parasitology, 91(2), 105-114 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service