Skip to Content
Merck
All Photos(3)

Key Documents

D131

Sigma-Aldrich

3,5-Dinitrocatechol

solid

Synonym(s):

3,5-Dinitro-1,2-benzenediol, OR-486

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N2O6
CAS Number:
Molecular Weight:
200.11
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

solid

color

yellow

mp

168-170  °C

solubility

H2O: slightly soluble 0.17 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 2.8 mg/mL
0.1 M HCl: slightly soluble
DMSO: soluble
aqueous buffer pH > 5: soluble
ethanol: soluble

storage temp.

2-8°C

SMILES string

Oc1cc(cc(c1O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O6/c9-5-2-3(7(11)12)1-4(6(5)10)8(13)14/h1-2,9-10H

InChI key

VDCDWNDTNSWDFJ-UHFFFAOYSA-N

Gene Information

human ... COMT(1312)

Related Categories

Application

3,5-Dinitrocatechol (3,5-DNC) has been used in the preparation of the molybdenum (VI)-(3,5-DNC) complex. It has also been used as a catechol-O-methyltransferase (COMT) inhibitor and as a positive control for screening human COMT inhibition.
Chelating reagent used in a sensitive (μM) assay for vanadium.

Biochem/physiol Actions

Selective inhibitor of catechol O-methyl transferase (COMT); penetrates the blood brain barrier and is useful both orally and parenteraly in experiments where inhibition of COMT in the central nervous system is required.

Quality

Solutions may be stored for several days at 4 °C.

Caution

Photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K Pani et al.
General pharmacology, 31(1), 67-73 (1998-05-22)
1. The efficacies of various agents that affect monoamine synthesis, oxidation and methylation were evaluated in the scallop, Placopecten magellanicus, through the use of high performance liquid chromatography with electrochemical detection. 2. Central ganglia, labial palps and feet from animals
V W Couling et al.
Biophysical journal, 75(2), 1097-1106 (1998-07-24)
This paper presents a study of the use of ultraviolet resonance Raman (UVRR) spectroscopic methods as a means of elucidating aspects of drug-protein interactions. Some of the RR vibrational bands of the aromatic amino acids tyrosine and tryptophan are sensitive
C Wiese et al.
Naunyn-Schmiedeberg's archives of pharmacology, 348(6), 582-585 (1993-12-01)
Organotypic primary cell cultures of fetal rat brain were used as a model system to study the effect of COMT inhibitors on the cerebral metabolic conversions of fluoro-DOPA enantiomers. The selective COMT inhibitors OR 486 and CGP 28014 were used
P T Männistö et al.
Life sciences, 43(18), 1465-1471 (1988-01-01)
Novel bisubstituted catechols were found to be potent and highly selective COMT inhibitors in vitro. One of them, OR-462 (3-(3,4-dihydroxy-5-benzylidene)-2,4-pentanedione), was studied also in vivo. When administered to rats orally together with levodopa and carbidopa, OR-462 greatly improved the bioavailability
Reda Saber Ibrahim Ahmed et al.
Journal of cellular biochemistry, 117(10), 2357-2369 (2016-03-08)
Uterine fibroids (leiomyomas) are very common benign tumors grown on the smooth muscle layer of the uterus, present in up to 75% of reproductive-age women and causing significant morbidity in a subset of this population. Although the etiology and biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service