D0447
D12-116 trifluoroacetate salt
≥98% (HPLC), powder
Synonym(s):
L-Tyrosyl-L-a-glutamyl-L-seryl-L-isoleucyl-L-arginyl-L-isoleucylglycyl-L-valyl-L-alanyl-L-prolyl-L-seryl-L-glutamine trifluoroacetate salt, YESIRIGVAPSQ trifluoroacetate salt
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to off-white
solubility
H2O: ≥2 mg/mL
storage temp.
−20°C
SMILES string
OC(=O)C(F)(F)F.CC[C@H](C)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](NC(=O)[C@H](CO)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](N)Cc1ccc(O)cc1)[C@@H](C)CC)C(=O)NCC(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C)C(=O)N2CCC[C@H]2C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(O)=O
InChI
1S/C58H94N16O19.C2HF3O2/c1-8-29(5)45(53(88)64-25-42(79)71-44(28(3)4)54(89)65-31(7)56(91)74-23-11-13-40(74)52(87)70-38(26-75)50(85)68-37(57(92)93)18-20-41(60)78)72-49(84)35(12-10-22-63-58(61)62)67-55(90)46(30(6)9-2)73-51(86)39(27-76)69-48(83)36(19-21-43(80)81)66-47(82)34(59)24-32-14-16-33(77)17-15-32;3-2(4,5)1(6)7/h14-17,28-31,34-40,44-46,75-77H,8-13,18-27,59H2,1-7H3,(H2,60,78)(H,64,88)(H,65,89)(H,66,82)(H,67,90)(H,68,85)(H,69,83)(H,70,87)(H,71,79)(H,72,84)(H,73,86)(H,80,81)(H,92,93)(H4,61,62,63);(H,6,7)/t29-,30-,31-,34-,35-,36-,37-,38-,39-,40-,44-,45-,46-;/m0./s1
InChI key
UVRSGFIIMHKCQD-ASTQSYHGSA-N
Biochem/physiol Actions
Features and Benefits
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service