Skip to Content
Merck
All Photos(1)

Key Documents

CRISPR07

Sigma-Aldrich

CRISPR UNIVERSAL NEGATIVE CONTROL 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

form

liquid

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Universal negative control CRISPRs have been designed not to recognize any sequence in the human, mouse or rat genome. A single vector format is provided which includes the Cas9 expression cassette and gRNA sequence. This vector includes GFP which is co-expressed from the same mRNA as the Cas9 protein via a 2A peptide linkage and enables for tracking of transfection efficiency or enrichment in cell populations via FACS.

Application

Functional Genomics/Target Validation
  • Creation of gene knockouts in cell lines
  • Creation of knock-in cell lines with promoters, fusion tags or reporters integrated into endogenous genes

Features and Benefits

Allows for expression of Cas9 and GFP without specific targeting of the CRISPR/Cas9 system.

Components

1 vial containing 1ug of U6-gRNA/CMV-Cas9-GFP plasmid expressing a non-targeting guide sequence.

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a single gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB.

Physical form

Sigma U6-gRNA/CMV-Cas9-GFP plasmid expressing a non-targeting guide sequence supplied at a concentration of 20ng/ul in 50ul.

Preparation Note

Sigma CRISPR plasmid products are delivered as mini-prep aliquots,
which may not be suitable for transfection into particular cell types. For best results, we advise maxi-prepping
plasmids using endotoxin-free DNA purification kits prior to transfection.

Other Notes

Typical transfection concentrations used in literature are in the ranges of 2.0 to 8.0ug of the single vector expressing the guideRNA and Cas9 . (All dosages above assume 0.5 to 1 million cells nucleofected)

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service