CLS6627
Corning® Step-R Repeating Pipettor Syringe Tips
volume (50.0 mL), non-sterile
Synonym(s):
pipet tips
All Photos(1)
About This Item
Recommended Products
sterility
non-sterile
packaging
pack of 25 ea
manufacturer/tradename
Corning 6627
volume
(50.0 mL)
General description
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)