CLS4975
Corning® aspiration pipettes
volume 1 mL
Synonym(s):
pipet
All Photos(1)
About This Item
UNSPSC Code:
41121500
eCl@ss:
32070201
NACRES:
NB.01
Recommended Products
material
polystyrene
sterility
sterile
manufacturer/tradename
Corning 4975
packaging
pkg of (50 per bag)
capacity
1 mL
volume
1 mL
Related Categories
General description
- Corning® aspirating pipettes are made of optically clear polystyrene for durability and good mechanical strength.
- The sterile, ungraduated, and unplugged pipettes are best suited for aspirating liquid aseptically using vacuum suction.
- Individually wrapped and bulk packed.
- Disposable and certified non-pyrogenic.
Aspirating pipettes are sterile, ungraduated and unplugged polystyrene pipettes for aspirating liquid using vacuum suction.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)