CLS4877
Corning® Transtar® liquid handling system
disposable reservoir liner, 12-channel
Synonym(s):
lab equipment accessory
All Photos(1)
About This Item
UNSPSC Code:
41121522
NACRES:
NB.11
Recommended Products
material
U-bottom bottom
white polystyrene
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
Corning 4877
volume measuring range
25-200 μL
capacity
200 μL
5 mL
volume
200 μL
Related Categories
General description
The Transtar is a portable, autoclavable liquid handling system designed for use with 96 well plates, A sterile 96 tip disposable cartridge, which loads into the Transtar system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 - 200 μL in 5 μL increments, Transtar accuracy us rated at 5% at all volume levels.
Legal Information
Corning is a registered trademark of Corning, Inc.
TRANSTAR-96 is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)