Skip to Content
Merck
All Photos(1)

Key Documents

CLS432083

Extension Collar, for Corning® CoolBox XT System

purple

Synonym(s):

Corning Coolcell accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123304
NACRES:
NB.12

packaging

pkg of 1

manufacturer/tradename

Corning 432083

L × W × H

14.0 cm × 9.4 cm × 7.0 cm, Internal Dimensions
20.0 cm × 16.0 cm × 7.0 cm, External Dimensions

color

purple

Looking for similar products? Visit Product Comparison Guide

General description

For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.

Legal Information

CoolBox is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service