CLS432005
Corning® CoolCell™ 5 mL LX Cell Freezing Container
purple
Synonym(s):
cryovials
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123300
NACRES:
NC.01
Recommended Products
material
foamed PE
packaging
pkg of 1
manufacturer/tradename
Corning 432005
technique(s)
cryopreservation: suitable
color
purple
Related Categories
General description
Corning® CoolCell™ freezer containers are ideal for cryopreservation of stem cells, primary cells, PBMC, cell lines, insect cells, yeast, and others. The proprietary Corning CoolCell technology utilizes a thermo-conductive alloy core and highly-insulative outer material to control the rate of heat removal and provide reproducible cell cryopreservation.
Features and Benefits
- Corning® CoolCell™, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute
- Easy to use with quick re-use time
- Inexpensive than alcohol-based devices and no ongoing cost involved
- Available in purple, green, orange, and pink
- Radially symmetric for all vials to have a uniform freeze rate
- Numbered wells for easy sample identificatio.
- Designed with a beveled lid for steady grip and easy opening
- Vial tips can be removed by tipping the device upside down
Legal Information
CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service