Skip to Content
Merck
All Photos(1)

Key Documents

CLS430157

Corning® culture tubes

15 mL culture tubes, 16 x 125 mm, w/ screw top plug seal cap, polystyrene, non-treated, sterile, 500/cs

Synonym(s):

cell culture tubes, plastic test tubes, plastic test tubes with caps, test tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121706
NACRES:
NB.13

material

cap (plugseal)
polystyrene
round bottom tube

sterility

sterile

feature

cap threaded; plug seal
disposable
Bottom style: round

packaging

pack of 50
case of 500

manufacturer/tradename

Corning 430157

capacity

15 mL

diam. × L

16 mm × 125 mm

color

clear

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Corning® cell culture tubes are manufactured from optically clear polystyrene.
  • Features a threaded plug seal cap to maximize sample recovery and prevent leakage.
  • Sterilized by gamma radiation and nonpyrogenic.
  • Non-treated surface is ideal for non-adherent cell culture.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service