Key Documents
CLS3300
Corning® 96 Well CellBIND® Microplates
flat bottom clear, polystyrene, sterile, lid
Synonym(s):
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, 96 well tissue culture treated plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear plate
flat bottom clear
flat bottom wells
polystyrene
polystyrene plate
round wells
size:
96 wells
material
clear plate
flat bottom clear
flat bottom wells
polystyrene
polystyrene plate
round wells
sterility
sterile
sterile
feature
lid
lid
skirt
plate format: 96 well standard
packaging
pack of 5
case of 50
manufacturer/tradename
Corning 3300
size
96 wells
surface area
0.32 cm2 , cell growth area
cell growth area
cell growth area
well volume
360 μL
well working volume
75-200 μL
suitability
suitable for (cell cuture)
binding type
CellBIND® surface
General description
  • Flat bottoms with 360 μL total volume
  • Recommended working volumes of 75 to 200 μL
  • CellBIND® surface treated for optimal cell attachment
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
Legal Information
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Yanli Ma et al.
PloS one, 7(10), e47961-e47961 (2012-11-01)
Aflatoxin B1 (AFB1) is a potent carcinogen that can induce hepatocellular carcinoma. AFB1-8,9-exo-epoxide, one of AFB1 metabolites, acts as a mutagen to react with DNA and induce gene mutations, including the tumor suppressor p53. In addition, AFB1 reportedly stimulates IGF