Skip to Content
Merck
All Photos(1)

Key Documents

CHSE-214 Cell Line from salmon

91041114, fish embryo, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Pricing and availability is not currently available.

product name

CHSE-214 Cell Line from salmon, 91041114, embryo

biological source

fish embryo

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Salmon embryo

Cell Line Description

Derived from a Chinook salmon (Oncorhynchus tshawytscha) embryo, susceptible to a wide range of fish viruses and in many instances replicate high titres.

Application

CHSE-214 cell line has been used for the growth and titration of infectious pancreatic necrosis virus (IPNV) from samples.[1] It has also been used to carry out cell infection assays to determine the effect of tropodithietic acid (TDA)-exposed isolates.[2]

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6; i.e seeding at 1-4 x 10,000 cells per cm using 0.25% trypsin or trypsin/EDTA, cells detach after 5-10 minutes at room temperature. Grow cells in dark (e.g. wrapped in foil); 5% CO2; 21°C. Freeze cells in 5% DMSO and 95% foetal bovine serum (FBS). Fish cell lines detach easily during transit if the culture is too young. For resuscitation seed flasks at 4-5 x 10,000 cells per cm and then split cells 1:3 to 1:4 and culture for at least one week with 1-2 media changes before shipping.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service