U937 Cell Line human
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85011440, human lung (lymphoblast), Lymphoblast
Synonym(s):
U-937 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
human lung (lymphoblast)
packaging
tube of 5 μg 85011440-DNA-5UG
pkg of vial of cells 85011440-1VL
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian histiocytic lymphoma
Cell Line Description
Derived from malignant cells of a pleural effusion of 37 year old caucasian male with diffuse histiocytic lymphoma. One of only a few human lines still expressing many of the monocytic like characteristics exhibited by cells of histiocytic origin.
Application
Tumourigenicity studies
U937 Cell Line human has been used to study the toxicity of propolis extracts against U937 cells and to study the uptake of apoptotic bodies (derived from monocytic U-937 cells) by primary meningothelial cells (PMECs).
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 10,12
D16S539: 12
D5S818: 12
D7S820: 9,11
THO1: 6,9.3
TPOX: 8,11
vWA: 14,15
CSF1PO: 12
D13S317: 10,12
D16S539: 12
D5S818: 12
D7S820: 9,11
THO1: 6,9.3
TPOX: 8,11
vWA: 14,15
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C. Cells may take up to 72 hours until confluent.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service