Skip to Content
Merck
All Photos(5)

Key Documents

C9002

Sigma-Aldrich

Hexadecylpyridinium chloride monohydrate

Synonym(s):

Cetylpyridinium chloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H38ClN · H2O
CAS Number:
Molecular Weight:
358.00
ISBN-10:
MFCD00149977
Beilstein:
3578606
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

technique(s)

DNA extraction: suitable

SMILES string

O.[Cl-].CCCCCCCCCCCCCCCC[n+]1ccccc1

InChI

1S/C21H38N.ClH.H2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-22-20-17-15-18-21-22;;/h15,17-18,20-21H,2-14,16,19H2,1H3;1H;1H2/q+1;;/p-1

InChI key

NFCRBQADEGXVDL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecylpyridinium chloride monohydrate is a cationic surfactant.

Application

Hexadecylpyridinium chloride monohydrate has been used in a study to assess removal of emulsified oil from oily waste water and removal of emulsified food and mineral oils from waste water.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shariff Ibrahim et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 62(5), 1177-1182 (2010-09-08)
An agricultural waste derived adsorbent was prepared by chemically modified barley straw with NaOH and a cationic surfactant hexadecylpyridinium chloride monohydrate (CPC). The prepared adsorbent, BMBS, was used for removal of anionic dyes; Acid Blue (AB40) and Reactive Blue 4
Removal of emulsified oil from oily wastewater using agricultural waste barley straw
Ibrahim, et al.
Biochemical Engineering Journal, 49, 6-6 (2010)
Wenxiang Chu et al.
Annals of translational medicine, 8(21), 1419-1419 (2020-12-15)
The removal of permanent internal fixation devices by secondary surgery could be avoided if these devices were made of degradable magnesium and magnesium alloys. Before such implants can be used clinically, however, the biological effect of magnesium exposure on surrounding
Removal of emulsified food and mineral oils from wastewater using surfactant modified barley straw
Ibrahim, S., et al.
Bioresource Technology, 100, 6-6 (2009)
Junkai Zeng et al.
Bioactive materials, 5(3), 435-446 (2020-04-14)
Osteoporosis bone defect is a refractory orthopaedic disease which characterized by impaired bone quality and bone regeneration capacity. Current therapies, including antiosteoporosis drugs and artificial bone grafts, are not always satisfactory. Herein, a strontium-substituted calcium phosphate silicate bioactive ceramic (Sr-CPS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service