Skip to Content
Merck
All Photos(4)

Key Documents

C8503

Sigma-Aldrich

Cholesterol

from sheep wool, ≥92.5% (GC), powder

Synonym(s):

3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Beilstein:
1915888
EC Number:
MDL number:
UNSPSC Code:
41141804
eCl@ss:
39023139
PubChem Substance ID:
NACRES:
NA.77

biological source

sheep wool

Assay

≥92.5% (GC)

form

powder

bp

360 °C (lit.)

mp

147-149 °C (lit.)

solubility

water: soluble 0.00003 g/L at 20 °C

density

1.067 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Cholesterol was used to prepare immune-stimulating complexes to enhance cellular and humoral responses. It was incorporated in animal feed to study the effect of cholesterol-rich diet.

Biochem/physiol Actions

Cholesterol is a lipid makes up about 20-25% of the structural components of the cell membranes. It determines the fluidity and permeability of the membrane, making it permeable to water but not to ions and protons. Cholesterol also regulates the functions of the transporters and signaling proteins present on the plasma membrane. The major sites of cholesterol synthesis are small intestine and liver.
Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of probiotic bacteria, isoflavones and simvastatin on lipid profile and atherosclerosis in cholesterol-fed rabbits: a randomized double-blind study
Cavallini DC et al
Lipids in Health and Disease, 8, doi: 10-doi: 10 (2009)
Effects of 2 or 5 consecutive exercise days on adipocyte area and lipid parameters in Wistar rats
Guerra RL et al
Lipids in Health and Disease, 6, doi: 10-doi: 10 (2007)
Anja Helgeby et al.
Journal of immunology (Baltimore, Md. : 1950), 176(6), 3697-3706 (2006-03-07)
The cholera toxin A1 (CTA1)-DD/QuilA-containing, immune-stimulating complex (ISCOM) vector is a rationally designed mucosal adjuvant that greatly potentiates humoral and cellular immune responses. It was developed to incorporate the distinctive properties of either adjuvant alone in a combination that exerted
John M Dietschy et al.
Journal of lipid research, 45(8), 1375-1397 (2004-07-16)
Unesterified cholesterol is an essential structural component of the plasma membrane of every cell. During evolution, this membrane came to play an additional, highly specialized role in the central nervous system (CNS) as the major architectural component of compact myelin.
R R Grummer et al.
Journal of animal science, 66(12), 3160-3173 (1988-12-01)
Cholesterol utilized for steroid synthesis by ovarian tissue may be derived from de novo synthesis or cellular uptake of lipoprotein cholesterol. The majority of blood cholesterol is transported by either low (LDL) or high (HDL) density lipoproteins, depending on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service