Skip to Content
Merck
All Photos(1)

Key Documents

C8145

Sigma-Aldrich

Cefsulodin sodium salt hydrate

third-generation cephalosporin antibiotic

Synonym(s):

7-(D-α-Sulfophenylacetamido)ceph-3-em-3-(4′-carbamoylpyridinium)methyl-4-carboxylate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N4NaO8S2 · xH2O
CAS Number:
Molecular Weight:
554.53 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
51284140
PubChem Substance ID:
NACRES:
NA.85

form

powder or crystals

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].NC(=O)c1cc[n+](CC2=C(N3[C@H](SC2)[C@H](NC(=O)[C@@H](c4ccccc4)S([O-])(=O)=O)C3=O)C([O-])=O)cc1

InChI

1S/C22H20N4O8S2.Na/c23-18(27)13-6-8-25(9-7-13)10-14-11-35-21-15(20(29)26(21)16(14)22(30)31)24-19(28)17(36(32,33)34)12-4-2-1-3-5-12;/h1-9,15,17,21H,10-11H2,(H4-,23,24,27,28,30,31,32,33,34);/q;+1/p-1/t15-,17-,21-;/m1./s1

InChI key

REACMANCWHKJSM-DWBVFMGKSA-M

General description

Chemical structure: ß-lactam

Application

Cefsulodin is a third generation cephalosporin antibiotic that has very specific activity against Pseudomonas aeruginosa. Cefuslodin is used in CIN (cefsulodin-Irgasan-novobiocin) agar to select for microorganisms such as E. coli. Cefsulodin is a substrate for rat Oatp1a4. It has been used to study multidrug resistance-associated protein 4 in efflux transport of prostaglandin E2 across the mouse blood-brain barrier.
Cefsulodin is used to study the effect of expression, binding, and inhibition of PBP1 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis.

Biochem/physiol Actions

Cefsulodin is a cephalosporin. Cephalosporins disrupt cell wall synthesis by inhibiting PBP crosslinking of peptidoglycan.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T H Tsai et al.
Journal of chromatography. A, 914(1-2), 83-87 (2001-05-19)
A microdialysis method followed by a microbore liquid chromatographic ultraviolet detection procedure has been performed for the assay of unbound cefsulodin in rat blood. A microdialysis probe was inserted into the jugular vein for blood sampling. This method involves an
Miguel Angel de Pedro et al.
Microbiology (Reading, England), 148(Pt 1), 79-86 (2002-01-10)
Periodic activation of zonal peptidoglycan (murein) synthesis at division sites in Escherichia coli has been reported recently. Zonal synthesis is responsible for septum formation, whereas elongation of the cell sacculus is performed by diffuse insertion of precursors. Zonal synthesis can
Friederike Hilbert et al.
International journal of food microbiology, 84(1), 111-115 (2003-06-05)
Yersinia enterocolitica is an important foodborne pathogen, but isolation of virulent Yersinia from food sources is still time consuming and requires skills. In this article, we describe a rapid urease screening on cefsulodin-irgasan-novobiocin (CIN) agar plates with an agar overlay
Danièle Joseleau-Petit et al.
Journal of bacteriology, 189(18), 6512-6520 (2007-06-26)
Growing bacterial L forms are reputed to lack peptidoglycan, although cell division is normally inseparable from septal peptidoglycan synthesis. To explore which cell division functions L forms use, we established a protocol for quantitatively converting a culture of a wild-type
M A Hornitzky et al.
FEMS microbiology letters, 198(1), 17-22 (2001-04-28)
The prevalence of complex Shiga toxin-producing Escherichia coli (STEC), i.e. STEC containing accessory virulence factors intimin (eaeA) and/or enterohaemorrhagic E. coli haemolysin (ehxA) and their serotypes were determined in diagnostic bovine faecal samples processed during a 3 months period. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service