Skip to Content
Merck
All Photos(4)

Documents

C8011

Sigma-Aldrich

Chelidamic acid hydrate

≥95%, powder

Synonym(s):

1,4-Dihydro-4-oxo-2,6-pyridinedicarboxylic acid hydrate, 4-Hydroxypyridine-2,6-dicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO5 · xH2O
CAS Number:
Molecular Weight:
183.12 (anhydrous basis)
Beilstein:
476229
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥95%

form

powder

color

brown

mp

267 °C (dec.) (lit.)

solubility

1 M NH4OH: 50 mg/mL, clear to slightly hazy

SMILES string

O.OC(=O)C1=CC(=O)C=C(N1)C(O)=O

InChI

1S/C7H5NO5.H2O/c9-3-1-4(6(10)11)8-5(2-3)7(12)13;/h1-2H,(H,8,9)(H,10,11)(H,12,13);1H2

InChI key

SNGPHFVJWBKEDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chelidamic acid hydrate has been used in eluent solution as a complexing additive in high-performance chelation ion chromatography (HPCIC) for speciation of iron. It has also been used as an organic ligand for the polyoxometalate (POM)-based lanthanoid hybrid compounds.

Biochem/physiol Actions

Chelidamic acid (CDA), a hydroxylated derivative of 2,6-pyridine-dicarboxylic acid (DPA), is a trianionic chelating agent.
Among the most potent of the tested "conformationally restricted glutamate analogs" as an inhibitor of glutamate decarboxylase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Construction of a dinuclear cluster containing La (?) and 4-hydroxypyridine-2, 6-dicarboxylic acid to modify Keggin-type polyoxometalate
Taghipour F and Mirzaei M
Journal of Molecular Structure null
Chelidamic acid monohydrate: the proton complex of a multidentate ligand
Hall et al.
Acta crystallographica. Section C, Crystal structure communications, 56 (Pt 4), 448-450 (2000-05-18)
Chelidamic acid as a new eluent for the determination of Fe (II) and Fe (III) species and other metals by high performance chelation ion chromatography
Dias J, et al.
Chromatographia null
Maryam Samaniyan et al.
Dalton transactions (Cambridge, England : 2003), 50(5), 1895-1900 (2021-01-22)
A novel inorganic-organic hybrid, {Na[Tb(L)(H2O)4]3(SiW12O40)}·4H2O, in which L = 4-hydroxypyridine-2,6-dicarboxylic acid (chelidamic acid), was prepared and characterized by elemental analyses, Fourier transform infrared spectroscopy, and single-crystal X-ray diffraction. It consists of a trinuclear terbium cluster bonded to a Keggin-type polyoxotungstate.
[Antiviral activity of chelidamic acid derivatives].
V E Iavorovskaia et al.
Voprosy virusologii, 29(3), 361-363 (1984-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service