Skip to Content
Merck
All Photos(4)

Key Documents

C7736

Sigma-Aldrich

Anti-Centrin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CEN1, Anti-CETN

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21 kDa

species reactivity

human

technique(s)

microarray: suitable
western blot: 1:2,000 using 293-T exp.Hu.Centrin-1LY117608 or 293-T exp.Hu.Centrin1-FLAG LY401316

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CETN1(1068)

General description

Centrins are acidic proteins, that are usually found in all eukaryotes. Centrin 1 (CETN1), an intron-less gene is mapped on human chromosome 18p11. It possesses certain features of an expressed retroposon.
The centrosome, the major microtubule-organizing center (MTOC) in the animal cell, is a key determinant in the organization of the cytoplasm, in the determination of cell polarity, and in the establishment of mitotic spindle poles during the cell cycle. Centrin is a member of the calcium binding E-F hand protein superfamily present in the centrosome pericentriolar matrix.
Three separate centrin genes have been identified in human cells: centrin 1, 2, and 3. Centrin 1 shows strong amino acid homology to centrin 2 (85%), but lower homology to centrin 3 (54%) and CDC31 (50%) from the yeast S. cerevisiae. Centrin has a fundamental role in the MTOC structure and function. Phenotypic analysis of mutants in Chlamydomonas caltractin and the yeast CDC31 genes have indicated that the protein is required for proper duplication and segregation of the MTOC in the respective cells.

Immunogen

synthetic peptide corresponding to the C-terminus of human centrin 1 (amino acids 157-172). The sequence is highly conserved in centrin 1 of many species, e.g. mouse, Drosophila, and Xenopus. It is also found in human and mouse centrin 2, but is not found in centrin 3.

Application

Anti-Centrin antibody produced in rabbit has been used in:
  • immunofluorescence
  • immunoblotting
  • immunofluorescence microscopy

Biochem/physiol Actions

Centrin has a fundamental role in the microtubule-organizing center (MTOC) structure and function. These proteins has imperative roles in centrosome positioning and segregation. Centrin also participates in cytokinesis and cell cycle progression.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clustering phenotype populations by genome-wide RNAi and multiparametric imaging
Fuchs F, et al.
Molecular Systems Biology, 6(1) (2010)
Optimization of purification method and characterization of recombinant human Centrin-1
Phanindranath R, et al.
Protein Expression and Purification, 124, 48-54 (2016)
Interleukin-13-Induced Mucous Metaplasia Increases Susceptibility of Human Airway Epithelium to Rhinovirus Infection
Lachowicz-SME, et al.
American Journal of Respiratory Cell and Molecular Biology (2012)
CETN1 is a cancer testis antigen with expression in prostate and pancreatic cancers
Kim JJ, et al.
Biomarker research, 1, 22-22 (2013)
Yuki Yoshino et al.
Journal of cell science, 133(17) (2020-08-14)
Breast cancer gene 1 (BRCA1) contributes to the regulation of centrosome number. We previously identified receptor for activated C kinase 1 (RACK1) as a BRCA1-interacting partner. RACK1, a scaffold protein that interacts with multiple proteins through its seven WD40 domains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service