Skip to Content
Merck
All Photos(1)

Key Documents

C5851

Sigma-Aldrich

CGP 35348 hydrate

≥97% (NMR), solid

Synonym(s):

(3-Aminopropyl)(diethoxymethyl)phosphinic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H20NO4P · xH2O
CAS Number:
Molecular Weight:
225.22 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (NMR)

form

solid

storage condition

desiccated

color

white

solubility

H2O: >20 mg/mL

originator

Novartis

storage temp.

−20°C

SMILES string

O.CCOC(OCC)P(O)(=O)CCCN

InChI

1S/C8H20NO4P.H2O/c1-3-12-8(13-4-2)14(10,11)7-5-6-9;/h8H,3-7,9H2,1-2H3,(H,10,11);1H2

InChI key

SRBHGEXMDCJOMS-UHFFFAOYSA-N

Application

CGP 35348 hydrate has been used as a γ-aminobutyric acid-B (GABAB) receptor inhibitor:
  • to study its effects on intrinsic optical signal (IOS) in rat hippocampus
  • alone or in combination with sodium salicylate to study its effects on auditory responses in the rat′s dorsal cortex of rat
  • to study its effects on hepatocellular carcinoma cells (HCC)

Biochem/physiol Actions

CGP 35348 is a γ-aminobutyric acid-B (GABAB) receptor antagonist. It has a higher affinity towards post-versus presynaptic receptors that can penetrate the blood-brain barrier.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAB Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Caution

Product is hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiyang Song et al.
Pain, 147(1-3), 241-248 (2009-10-20)
The aim of the present study was to examine the role of the spinal serotonergic system in the pain relieving effect of spinal cord stimulation (SCS) using a rat model of mononeuropathy. Tactile withdrawal thresholds, cold responses and heat withdrawal
Abigail J Houston et al.
European journal of pharmacology, 674(2-3), 327-331 (2011-12-07)
The effects of subcutaneous (s.c.) administration of the GABA(B) receptor agonist baclofen were investigated on primary drinking in rats. Baclofen (1-4 mg/kg) produced a dose-related reduction in cumulative water intake in 16 h water-deprived rats during the 120 min measurement
Madhu P Sirivelu et al.
Brain research, 1248, 107-114 (2008-11-19)
Interleukin-1beta (IL-1beta), a cytokine that is closely associated with inflammation and immune stress, is known to interfere with reproductive functions. Earlier studies have demonstrated that IL-1beta inhibits the luteinizing hormone (LH) surge during the afternoon of proestrus in female rats.
Pavel Mareš
Epilepsy research, 92(2-3), 125-133 (2010-09-25)
Epileptic seizures activate not only excitatory but also inhibitory systems what results in an arrest of seizures. Our recent data indicate that GABAergic inhibition plays an important role in this process in cortically elicited seizures. We started to study the
S E Sander et al.
Neuroscience, 162(2), 423-430 (2009-05-12)
The GABA(B) receptor (GABA(B)R) agonist baclofen is known to have a beneficial potency in patients who suffer from dystonia, a neurological syndrome characterized by involuntary co-contractions of opposing muscles. The underlying mechanisms of this movement disorder are still unclear. Previous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service