Skip to Content
Merck
All Photos(5)

Key Documents

C4290

Sigma-Aldrich

Butyrylcholinesterase from equine serum

lyophilized powder, ≥500 units/mg protein

Synonym(s):

Acylcholine acyl-hydrolase, Choline esterase, butyryl, Pseudocholinesterase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

equine serum

Quality Level

form

lyophilized powder

specific activity

≥500 units/mg protein

composition

Protein, ≥10%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Butyrylcholinesterase (BChE) from equine serum has been used:
  • to determine the inhibitory concentration of bupivacaine on butyrylcholinesterase[1]
  • in acetylcholinesterase (AChE)/BChE activity assay to determine the inhibitory activity of benzothiazole-piperazine compounds[2]

Selective inhibition of BChE activity can be used in the detection of organophosphates.[3] Its use in the treatment of organophosphate toxicity shows promise. There is a correlation between the level of BChE in human blood and degree of protection against potentially toxic nerve agents.[4] There has also been interest in the roles of cholinesterases with regard to Alzheimer′s disease. Investigations into selective inhibitors may provide a clearer picture of the physiological role of BChE in both healthy and diseased individuals.[5] The enzyme has been used to test bupivacaine as an inhibitor of butyrylcholinesterase during acetylcholinesterase assay using cerebrospinal fluid.[6]

Biochem/physiol Actions

Butyrylcholinesterase (BChE) is a serine hydrolase that is structurally similar to acetylcholinesterase (AChE), but differs in substrate specificities and inhibitor sensitivities. BChE can, unlike AChE, efficiently hydrolyze larger esters of choline such as butyrylcholine and benzoylcholine. The enzyme is a tetrameric glycoprotein with four equal subunits (110 kDa each). The enzyme is activated by Ca2+ and Mg2+ and the activity is constant over the pH range 6.0-8.0. It is inhibited by Betaine, nicotine, organophosphates, carbamates.

Unit Definition

One unit will hydrolyze 1.0 μmole of butyrylcholine to choline and butyrate per min at pH 8.0 at 37 °C. The activity obtained using butyrylcholine as substrate is ~2.5 times that obtained using acetylcholine.

Physical form

Highly purified, lyophilized powder containing buffer salts

Analysis Note

Protein determined by biuret

inhibitor

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Cinti et al.
Nature protocols, 14(8), 2437-2451 (2019-07-05)
Despite substantial advances in sensing technologies, the development, preparation, and use of self-testing devices is still confined to specialist laboratories and users. Decentralized analytical devices will enormously impact daily lives, enabling people to analyze diverse clinical, environmental, and food samples
Determination of thyroxine binding globulin.
Bergmeyer, H.U
Methods of Enzymatic Analysis, 2, 833-833 (1974)
W H Kluge et al.
BMC biochemistry, 2, 17-17 (2002-01-22)
Most test systems for acetylcholinesterase activity (E.C.3.1.1.7.) are using toxic inhibitors (BW284c51 and iso-OMPA) to distinguish the enzyme from butyrylcholinesterase (E.C.3.1.1.8.) which occurs simultaneously in the cerebrospinal fluid. Applying Ellman's colorimetric method, we were looking for a non-toxic inhibitor to
Luisa Savini et al.
Journal of medicinal chemistry, 46(1), 1-4 (2002-12-28)
Tacrine-based AChE and BuChE inhibitors were designed by investigating the topology of the active site gorge of the two enzymes. The homobivalent ligands characterized by a nitrogen-bridged atom at the tether level could be considered among the most potent and
Acetylcholinesterase assay for cerebrospinal fluid using bupivacaine to inhibit butyrylcholinesterase
Kluge WH, et al.
BMC Biochemistry, 2(1), 17-17 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service