Skip to Content
Merck
All Photos(1)

Key Documents

C-39606

Sigma-Aldrich

Renal Epithelial Cell Growth Medium 2

SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml

Synonym(s):

Recommended with C-26230

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

aqueous solution

packaging

pkg of 1 vial

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

Renal Epithelial Cell Growth Medium 2 SupplementMix (basal medium sold separately)

To form a complete medium, it should be combined with the basal medium: C-26230

Application

Renal Epithelial Cell Growth Medium 2 is an optimized low-serum (5% V/V) medium that additionally contains Epinephrine. The combination of low serum and selected growth factors ensures successful growth in vitro making the Renal Epithelial Cell Growth Medium 2 the first choice for PromoCell Renal Epithelial Cells. The Renal Epithelial Cell Growth Medium 2 is optimized for primary human cells, but can also be used for porcine and mouse renal epithelial cells.Recommended for:Human Renal Epithelial Cells (HREpC)Human Renal Cortical Epithelial Cells (HRCEpC)

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Renal Epithelial Cell Media are subjected to comprehensive quality control tests using primary human renal epithelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested renal epithelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.05 ml / ml Epidermal Growth Factor (recombinant human): 10 ng / mlInsulin (recombinant human): 5 µg / mlHydrocortisone: 36 ng / mlEpinephrine: 0.5 µg / mlTriiodo-L-thyronine: 4 pg / mlTransferrin (recombinant human): 5 µg / ml

Storage and Stability

Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

recommended

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service