C3821
Citrate-dextrose solution (ACD)
sterile-filtered
Synonym(s):
Acid citrate-dextrose, Anticoagulant citrate-dextrose
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Related Categories
General description
Citrate-dextrose solution (ACD) is an anticoagulant and preservative used routinely for blood storage. The ideal red blood cells shelf life is 21 days when stored in ACD solution. Red blood cells in ACD solution utilize glucose and their viability is impairs when reinfused.
Application
Citrate-dextrose solution (ACD) has been used for dilution of blood for haematological studies. It has also been used as an anticoagulant during blood collection by cardiac puncture from mice and human bone marrow isolation.
For research use only. Not for use in diagnostic procedures.
Biochem/physiol Actions
Useful for anticoagulation of whole blood; erythrocyte survival
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Isolation, expansion and characterisation of mesenchymal stem cells from human bone marrow, adipose tissue, umbilical cord blood and matrix: a comparative study
Secunda R, et al.
Cytotechnology, 67(5), 793-807 (2015)
Mingming Yang et al.
Experimental and therapeutic medicine, 20(2), 1253-1260 (2020-08-09)
The p53 gene is well known as a key tumor suppressor gene; it is vital for hematopoietic stem cell differentiation and growth. In the present study, the change of platelets (PLTs) in p53 knockout mice (p53-/- mice) was investigated. The
Interaction between FOG-1 and the corepressor C-terminal binding protein is dispensable for normal erythropoiesis in vivo
Katz SG, et al.
Molecular and Cellular Biology, 22(9), 3121-3128 (2002)
Hideki Miyachi et al.
International journal of cardiology, 266, 61-66 (2018-06-12)
Acute thrombosis is a crucial cause of bioresorbable vascular graft (BVG) failure. Bone marrow-derived mononuclear cell (BM-MNC)-seeded BVGs demonstrated high graft patency, however, the effect of seeded BM-MNCs against thrombosis remains to be elucidated. Thus, we evaluated an antithrombotic effect
The storage of hard-packed red blood cells in citrate-phosphate-dextrose (CPD) and CPD-adenine (CPDA-1)
Beutler E and West C
Blood, 54(1), 280-284 (1979)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service