Skip to Content
Merck
All Photos(2)

Key Documents

C2832

Sigma-Aldrich

Cholesteryl 3β-N-(di­methyl­amino­ethyl)­carbamate hydrochloride

≥95%

Synonym(s):

3β-[N-(Dimethylaminoethane)­carbamoyl]­cholesterol, DC-Chol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H57ClN2O2
CAS Number:
Molecular Weight:
537.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

Cl[H].[H][C@@]1(CC[C@@]2([H])[C@]3([H])CC=C4C[C@H](CC[C@]4(C)[C@@]3([H])CC[C@]12C)OC(=O)NCCN(C)C)[C@H](C)CCCC(C)C

InChI

1S/C32H56N2O2.ClH/c1-22(2)9-8-10-23(3)27-13-14-28-26-12-11-24-21-25(36-30(35)33-19-20-34(6)7)15-17-31(24,4)29(26)16-18-32(27,28)5;/h11,22-23,25-29H,8-10,12-21H2,1-7H3,(H,33,35);1H/t23-,25+,26+,27-,28+,29+,31+,32-;/m1./s1

InChI key

ISXSJGHXHUZXNF-LXZPIJOJSA-N

Application

Cholesteryl 3β-N-(di­methyl­amino­ethyl)­carbamate hydrochloride has been used as a bilayer-membrane stabiliser and for nucleic acid transfer.
This cationic cholesterol derivative forms liposomes with L-α-phosphatidylethanolamine, dioleoyl, that efficiently transfect mammalian cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene Targeting Protocols (2000)
S Szala et al.
Gene therapy, 3(11), 1026-1031 (1996-11-01)
An attempt was made to use simple cationic liposomes DC-Chol/DOPE and DDAB/DOPE (DC-Chol is 3 beta (N(N',N-dimethylaminoethane) carbamoyl) cholesterol, DDAB is dimethyldioctadecyl ammonium bromide and DOPE is dioleoylphosphatidylethanolamine) for transfer of Escherichia coli cytosine deaminase 'suicide' gene under the control
M J Campbell
BioTechniques, 18(6), 1027-1032 (1995-06-01)
Cationic liposomes are being utilized for the delivery of DNA into mammalian cells both in vitro and in vivo. In this report, we describe a rapid, simple injection method for the preparation of cationic liposomes that requires no special equipment.
A Colosimo et al.
Biochimica et biophysica acta, 1419(2), 186-194 (1999-07-17)
We evaluated the transfection efficiency of five different cationic liposome/plasmid DNA complexes, during the in vitro gene transfer into human epithelial tracheal cell lines. A dramatic correlation between the transfection efficiency and the charge ratio (positive charge of liposome to
N J Caplen et al.
Gene therapy, 2(9), 603-613 (1995-11-01)
Clinical trials using cationic liposome-mediated DNA transfer have now been initiated for several disorders including cystic fibrosis. Previous studies have shown that the level of gene expression achieved may be dependent on the formulation of the DNA-liposome complex and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service