C-27438
Adipocyte Nutrition Medium
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Adipocyte Nutrition Medium (Ready-to-use)
Application
Adipocyte Nutrition Medium can be used to cultivate mature adipocytes. The medium is optimized for primary human cells but can also be used for murine adipocytes.Recommended for:Human Adipocytes
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials
Quality
All lots of PromoCell Preadipocyte/Adipocyte Media are subjected to comprehensive quality control tests using primary human white preadipocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested preadipocytes. In addition, the Preadipocyte Differentiation Medium/Adipocyte Nutrition Medium are tested for the capacity to induce the terminal differentiation of preadipocytes into mature adipocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Preparation Note
Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.03 ml / mld-Biotin: 8 µg / mlInsulin (recombinant human): 0.5 µg / mlDexamethasone: 400 ng / ml
Storage and Stability
Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix or SupplementPack at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C
Subculture Routine
Click here for more information.
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service