Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NB.22
UNSPSC Code:
41103025
Manufacturer/tradename:
Nalgene 5115-0012
Feature:
capacity 12 vial(s), for tubes 0.2 ‑ 0.5 mL
Packaging:
pack of 1 ea
Pricing and availability is not currently available.
feature
capacity 12 vial(s), for tubes 0.2 ‑ 0.5 mL
packaging
pack of 1 ea
manufacturer/tradename
Nalgene 5115-0012
L × W × H
151 mm × 108 mm × 125 mm
weight
1.5 lb
General description
Maintains 12 microtubes below −15°C for up to 1 hr at room temperature.
Molded from durable polycarbonate and filled with non-toxic freezable gel. A convenient alternative to ice buckets for keeping reagents and enzymes cool. Also protects critical samples from temperature fluctuations in the freezer.
Domed lid permits storage of taller screw cap microtubes. Lids have gridded write-on surfaces for sample identification. Stackable.
Domed lid permits storage of taller screw cap microtubes. Lids have gridded write-on surfaces for sample identification. Stackable.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
1 of 1
This Item | |||
|---|---|---|---|
| feature capacity 12 vial(s), for tubes 0.2 ‑ 0.5 mL | feature Holds cryognic vials and 0.2 to 0.5mL microcentrifuge tubes | feature Holds cryognic vials and 0.2 to 0.5mL microcentrifuge tubes | feature - |
| manufacturer/tradename Nalgene 5115-0012 | manufacturer/tradename Nalgene® DS5116-0012 | manufacturer/tradename Nalgene® DS5116-0032 | manufacturer/tradename Nalgene® 5116-1300 |
| packaging pack of 1 ea | packaging - | packaging - | packaging pack of 1 ea |
| weight 1.5 lb | weight - | weight - | weight - |
| L × W × H 151 mm × 108 mm × 125 mm | L × W × H 151 mm × 108 mm × 125 mm | L × W × H 243 mm × 157 mm × 146 mm | L × W × H 197 mm × 140 mm × 190 mm |
Still not finding the right product?
Explore all of our products under Nalgene® Labtop cooler
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
