C-23060
Skeletal Muscle Cell Growth Medium
Ready-to-use kit including Basal Medium and SupplementMix, 500 ml
Synonym(s):
Primary Cell Culture Media
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.81
Recommended Products
form
liquid
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Skeletal Muscle Cell Growth Medium (Ready-to-use)
Application
Skeletal Muscle Cell Growth Medium is a low-serum medium (5% V/V) developed for the expansion of human skeletal muscle cell cultures. The media are optimized for primary human cells, but can also be used for murine, rat, canine, and rabbit skeletal muscle cells.Recommended for:Human Skeletal Muscle Cells (SkMC)
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials
Quality
All lots of PromoCell Skeletal Muscle Cell Growth Medium are subjected to comprehensive quality control tests using human primary SkMC. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested SkMC. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Preparation Note
Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.05 ml / mlFetuin (bovine): 50 µg / mlEpidermal Growth Factor (recombinant human): 10 ng / mlBasic Fibroblast Growth Factor (recombinant human): 1 ng / mlInsulin (recombinant human): 10 µg / mlDexamethasone: 0.4 µg / ml
Storage and Stability
Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix or SupplementPack at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Subculture Routine
Click here for more information.
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service