C-21160
Airway Epithelial Cell Growth Medium
Kit including Basal Medium and SupplementPack, 500 ml
Synonym(s):
Airway Cell Medium, Epithelial Growth Medium, Growth Medium for Airway Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
packaging
pkg of 500 ml
technique(s)
cell culture | mammalian: suitable
shipped in
wet ice
General description
Airway Epithelial Cell Growth Medium Kit
Application
Airway Epithelial Cell Growth Medium is a serum-free medium optimized for the in vitro cultivation of epithelial cells from large air passages. The medium is optimized for primary human cells, but can also be used for equine, porcine, murine, bat and rat airway epithelial cells.
Recommended for:
Human Nasal Epithelial Cells (HNEpC)
Human Bronchial Epithelial Cells (HBEpC)
Human Tracheal Epithelial Cells (HTEpC)
Recommended for:
Human Nasal Epithelial Cells (HNEpC)
Human Bronchial Epithelial Cells (HBEpC)
Human Tracheal Epithelial Cells (HTEpC)
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials
Quality
All lots of PromoCell Airway Epithelial Cell Growth Medium are subjected to comprehensive quality control tests using primary human airway epithelial cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested airway epithelial cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Preparation Note
Final supplement concentrations after addition to the basal medium:Bovine Pituitary Extract: 0.004 ml / ml Epidermal Growth Factor (recombinant human): 10 ng / mlInsulin (recombinant human): 5 µg / mlHydrocortisone: 0.5 µg / mlEpinephrine: 0.5 µg / mlTriiodo-L-thyronine: 6.7 ng / mlTransferrin, holo (human): 10 µg / mlRetinoic Acid: 0.1 ng / ml
Storage and Stability
Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix or SupplementPack at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, prewarm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Subculture Routine
Click here for more information.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service