Skip to Content
Merck
All Photos(3)

Key Documents

C1603

Sigma-Aldrich

Anti-Calcium Channel (α1C Subunit) (L-type of Voltage-gated Ca2+ Channel) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CACH2, Anti-CACN2, Anti-CACNL1A1, Anti-CCHL1A1, Anti-CaV1.2, Anti-LQT8, Anti-TS, Anti-TS. LQT8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse, rabbit

technique(s)

immunohistochemistry: suitable using Rat brain sections
western blot: 1:200 using Rat brain and rat heart membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Cacna1c(12288)

Related Categories

General description

L-type of Voltage-gated Ca2+ Channels (L-type VGCC) is a high voltage-activated channel that exerts spatial and temporal control over cellular calcium concentrations and modulates function such as neurotransmitter release, hormone secretion, muscle contraction, electrical activity, cell metabolism and proliferation, gene expression and neuronal survival. Calcium dependent processes are important in synaptic modification and thus alterations in calcium channel function may be important for both modifying synaptic plasticity and also in age-related neurodegenerative diseases. Calcium channel antagonists are used as antiarrhythmics and in the treatment of hypertension and may even be neuroprotective in Parkinson′s Disease. Calcium Channel α1C subunit is a pore-forming subunit that together with β subunits generates the necessary signal for membrane targeting of class C L-type calcium channels.

Specificity

Anti-Voltage-gated Calcium Channel α1C subunit recognizes the α1C subunit by immunoblotting in rat brain and rat heart membranes. Reactivity has been confirmed for rat, mouse, rabbit and human.

Immunogen

synthetic peptide corresponding to amino acids 848 to 865 of rat α1C.

Application

Anti-Calcium Channel (α1C Subunit) (L-type of Voltage-gated Ca2+ Channel) antibody produced in rabbit has been used in:
  • SDS-PAGE
  • western blotting
  • coimmunoprecipitation
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Rabbit polyclonal anti-Calcium Channel α1C subunit antibody is used to tag class C L-type calcium channel subunit α1C for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques such as ELISA, immunoblotting, and immunoprecipitation. It is used as a probe to determine the presence and roles of calcium channel subunit α1C in class C L-type voltage-gated Ca2+ channel subunit complexes.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Functional coupling of the metabotropic glutamate receptor, InsP3 receptor and L-type Ca2+ channel in mouse CA1 pyramidal cells
Kato HK, et al.
The Journal of Physiology, 590(13), 3019-3034 (2012)
Calcium channel function and regulation in beta1-and beta2-adrenoceptor transgenic mice
Foerster K, et al.
Naunyn-Schmiedeberg'S Archives of Pharmacology, 369(5), 490-495 (2004)
Nina Daschil et al.
Pharmacology, 93(1-2), 24-31 (2014-01-18)
It is well established that reactive astrocytes express L-type calcium channels (LTCC), but their functional role is completely unknown. We have recently shown that reactive astrocytes highly express the CaV1.2 α1-subunit around β-amyloid (Aβ) plaques in an Alzheimer mouse model.
Jing Liu et al.
American journal of physiology. Cell physiology, 297(1), C152-C159 (2009-04-10)
Cardiomyocytes (CMs) are nonregenerative. Self-renewable pluripotent human embryonic stem cells (hESCs) can differentiate into CMs for cell-based therapies. We recently reported that Ca(2+) handling, crucial to excitation-contraction coupling of hESC-derived CMs (hESC-CMs), is functional but immature. Such immature properties as
A S Axelsson et al.
Nature communications, 8, 15652-15652 (2017-06-07)
Type 2 diabetes (T2D) is characterized by insulin resistance and impaired insulin secretion, but the mechanisms underlying insulin secretion failure are not completely understood. Here, we show that a set of co-expressed genes, which is enriched for genes with islet-selective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service