Skip to Content
Merck
All Photos(1)

Key Documents

C1108

Sigma-Aldrich

Anti-Cannabinoid Receptor 1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CANN6, Anti-CB-R, Anti-CB1, Anti-CB1A, Anti-CB1K5, Anti-CB1R, Anti-CNR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen ~52 kDa

species reactivity

rat, human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CNR1(1268)
rat ... Cnr1(25248)

Looking for similar products? Visit Product Comparison Guide

Immunogen

fusion protein containing amino acids 1-99 of the human CB1 receptor.

Application

Anti-Cannabinoid Receptor 1 antibody produced in rabbit is suitable for western blotting at a working dilution of 1:250 and for immunocytochemistry at a working dilution of 1:1000.

Biochem/physiol Actions

Cannabinoids exert their well-known physiological effects through two G-protein coupled receptors, CB1 and CB2. The cannabinoid receptor type 1 (CB1) is widely distributed in mammalian tissues, with the highest concentrations found in brain neurons while CB2 receptor function primarily in the peripheral immune system, found in spleen and may be present in the central nervous system, including the retina. The cannabinoid receptor type 1 are involved in modulation of adenylate cyclase and ion channels. It is also involved in the modulation of nociception, movement, learning and memory, emotion and many other neuronal processes. It also plays an essential role in drug addiction. CB1 receptor blockade may be useful for treatment of angiotensin II-dependent hypertension and associated metabolic syndrome.

Physical form

Solution in phosphate buffered saline containing 1 mg/ml bovine serum albumin, 50% glycerol, and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hajar Miranzadeh Mahabadi et al.
Molecular brain, 14(1), 89-89 (2021-06-09)
The type 1 and type 2 cannabinoid receptors (CB1 and CB2 receptors) are class A G protein-coupled receptors (GPCRs) that are activated by endogenous lipids called endocannabinoids to modulate neuronal excitability and synaptic transmission in neurons throughout the central nervous
R G Pertwee
Current medicinal chemistry, 6(8), 635-664 (1999-09-02)
Mammalian tissues contain at least two types of cannabinoid receptor, CB1 and CB2, both coupled to G proteins. CB1 receptors are expressed mainly by neurones of the central and peripheral nervous system whereas CB2 receptors occur in certain non-neuronal tissues
Leyre Echeazarra et al.
Histochemistry and cell biology, 156(5), 479-502 (2021-08-29)
Specific and selective anti-CB1 antibodies are among the most powerful research tools to unravel the complex biological processes mediated by the CB1 receptor in both physiological and pathological conditions. However, low performance of antibodies remains a major source of inconsistency
C C Felder et al.
Annual review of pharmacology and toxicology, 38, 179-200 (1998-05-23)
Marijuana has been in use for over 4000 years as a therapeutic and as a recreational drug. Within the past decade, two cannabinoid receptor types have been identified, their signal transduction characterized, and an endogenous lipid agonist isolated from mammalian
Thangesweran Ayakannu et al.
Frontiers in oncology, 9, 430-430 (2019-06-28)
Objective: To identify new biochemical markers for endometrial cancer (EC). Recent evidence suggests that members of the endocannabinoid system (N-acylethanolamines) that bind to and activate receptors that are dysregulated in EC are involved in this tumour's biology. These observations suggest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service