Skip to Content
Merck
All Photos(1)

Key Documents

C0858

Sigma-Aldrich

Cyclooxygenase 2 human

≥70% (SDS-PAGE), recombinant, expressed in insect cells, aqueous solution, ≥8000 units/mg protein

Synonym(s):

COX-2, Prostaglandin H synthase 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in insect cells Active recombinant N-terminal His-tagged COX-2 N580A mutant purified from insect cells.
expressed in insect cells

Quality Level

Assay

≥70% (SDS-PAGE)

form

aqueous solution

specific activity

≥8000 units/mg protein

mol wt

dimer subunit mol wt 70-74 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTGS2(5743)

Biochem/physiol Actions

COX-2 catalyzes the conversion of arachidonic acid to prostaglandin H2 (the first step in the biosynthesis of prostaglandins, thromboxanes, and prostacyclins). It is the inducible form of cyclooxygenase, induced by phorbol esters, lipopolysaccharides, and cytokines. COX-2 is upregulated in the neovasculature of malignant tumors. Inhibition of COX-2 with nonsteroidal anti-inflammatory drugs (NSAIDs) is associated with decreased tumor growth.

Other Notes

Tagged with a six-residue histidine sequence near the N-terminus.

Unit Definition

One unit will consume 1.0 nanomole of oxygen per minute at pH 8 at 37 °C.

Physical form

Solution in 80 mM Tris, pH 8.0, with 0.3 mM DDC, 0.01% TWEEN® 20 and 10% glycerol

Substrates

KM for arachidonate = 6.5 μM. Exhibits activity and sensitivity to NSAIDs similar to the non-tagged enzyme.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandra Moreno et al.
Natural product communications, 10(6), 991-994 (2015-07-23)
Zuccagnia punctata Cav. (Fabaceae) is an Argentine medicinal aromatic shrub (jarilla pispito, puspus, lata and jarilla macho). The chalcones were identified as pigments responsible for the yellow color of the flowers. Hydroethanolic extracts were obtained both from fresh flowers and
Masanori Sugimoto et al.
Drug development research, 77(1), 20-28 (2016-01-15)
Preclinical Research This article describes the properties of a novel topical NSAID (Nonsteroidal anti-inflammatory drug) patch, SFPP (S(+)-flurbiprofen plaster), containing the potent cyclooxygenase (COX) inhibitor, S(+)-flurbiprofen (SFP). The present studies were conducted to confirm human COX inhibition and absorption of
Mater H Mahnashi et al.
Evidence-based complementary and alternative medicine : eCAM, 2022, 7921408-7921408 (2022-04-12)
Background. The current study aims to give a scientific origin for employing Habenaria plantaginea Lindl. as a potential candidate against nociception, inflammation, and pyrexia. The pharmacological studies were performed on crude extract and subfractions. In the gas chromatography-mass spectroscopy analysis
María J Pérez et al.
Journal of food science, 83(5), 1454-1462 (2018-04-17)
This work is part of the search in native food matrices from arid regions of Argentina of interest to improve human health. Prosopis species are ethnic food resources in South America capable of growing in arid and semi-arid environments. This
Muhammad Saeed Jan et al.
European journal of medicinal chemistry, 186, 111863-111863 (2019-11-20)
In recent years, drug discovery paradigm has been shifted from conventional single target inhibition toward multitarget design concept. In current research, we have reported synthesis, in-vitro, in-vivo and acute toxicity determination of N-substituted pyrrolidine-2,5-dione derivatives as multitarget anti-inflammatory agents. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service