Skip to Content
Merck
All Photos(8)

Key Documents

C0768

Sigma-Aldrich

Cyclophosphamide monohydrate

bulk package

Synonym(s):

2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide, Cytoxan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15Cl2N2O2P · H2O
CAS Number:
Molecular Weight:
279.10
Beilstein:
4678992
EC Number:
MDL number:
UNSPSC Code:
51112507
PubChem Substance ID:
NACRES:
NA.77

Assay

97.0-103.0% (HPLC)

form

powder

mp

49-51 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]O[H].ClCCN(CCCl)P1(=O)NCCCO1

InChI

1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2

InChI key

PWOQRKCAHTVFLB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Cyclophosphamide (CYC) is a cytotoxic alkylating agent. It is activated by the cytochrome P-450 enzyme system in the liver. It has immunomodulatory functionality and inhibits humoral and cellular immunity.Cyclophosphamide exhibits anti-cancer and immunosuppressant activities. In the body, cyclophosphamide is converted by the liver to an active alkylating 4-hydroxycyclophosphamide metabolite, thereby showing its anti-neoplastic effects.

Application

Cyclophosphamide monohydrate has been used:
  • to test its antitumor effect on TC-1 tumor cells
  • as a component of multidrug solution for isolation of resistant human burkitt lymphoma cell line
  • in testing antitumor immunity in mouse tumor cell lines

Biochem/physiol Actions

Cyclophosphamide is a cytotoxic nitrogen mustard derivative widely used in cancer chemotherapy. It cross-links DNA, causes strand breakage, and induces mutations. Its clinical activity is associated with a decrease in aldehyde dehydrogenase 1 (ALDH1) activity.
Cytotoxic nitrogen mustard derivative widely used in cancer chemotherapy. It cross-links DNA, causes strand breakage, and induces mutations.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Muta. 1B - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclophosphamide pharmacokinetics and dose requirements in patients with renal insufficiency
Haubitz M, et al.
Kidney International, 61(4), 1495-1501 (2002)
Cyclophosphamide and cancer: golden anniversary
Emadi A, et al.
Nature Reviews. Clinical Oncology, 6(11), 638-638 (2009)
Metronomic cyclophosphamide activation of anti-tumor immunity: tumor model, mouse host, and drug schedule dependence of gene responses and their upstream regulators
Wu J, et al.
BMC Cancer, 16(1), 623-623 (2016)
Milly E de Jonge et al.
Clinical pharmacokinetics, 44(11), 1135-1164 (2005-10-20)
Cyclophosphamide is an extensively used anticancer and immunosuppressive agent. It is a prodrug undergoing a complicated process of metabolic activation and inactivation. Technical difficulties in the accurate determination of the cyclophosphamide metabolites have long hampered the assessment of the clinical
J Tammam et al.
British journal of pharmacology, 158(5), 1183-1195 (2009-09-25)
gamma-Secretase inhibitors (GSIs) block NOTCH receptor cleavage and pathway activation and have been under clinical evaluation for the treatment of malignancies such as T-cell acute lymphoblastic leukaemia (T-ALL). The ability of GSIs to decrease T-ALL cell viability in vitro is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service