BR780546
BRAND® microcentrifuge tube, 2 mL with lid, PP
clear
Synonym(s):
Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
sterility
non-sterile
feature
RNase and DNase free
endotoxin free
manufacturer/tradename
BRAND 780546
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- BRAND® microcentrifuge tube is made from polypropylene (PP).
- Tubes are free from RNase, DNase, and endotoxins.
- Withstands RCF up to 20,000 × g at 20°C for 20 minutes.
- Graduations on one side with frosted marking area on the other side.
- Features a lid to protect samples from evaporation.
- Lid membrane with a 0.28 mm piercing area.
Features and Benefits
- Matte label field and division for rough volume reading
- Disposable microtubes
- Contains cover lock.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service