Key Documents
BAF133550001
Fluorescent Ruler, Metric/English Scale
pink phosphorescent ink visible under UV
About This Item
UNSPSC Code:
41106000
NACRES:
NB.22
Pricing and availability is not currently available.
manufacturer/tradename
Bel-Art F13355-0001
measuring range
0-15 cm
General description
This plastic metric/English scale is printed with a highly visible, glow-in-the-dark pink phosphorescent ink. Under UV light the ruler graduations glow visibly and help to calculate nucleic acid mobility. The ruler is marked 0 to 15cm in millimeter increment, along one edge and 0" to 6" in 1/16" increments on the other edge.
High visibility, glow-in-the-dark scale
  • Ideal for viewing fluorescent images and fluorescent photography