Skip to Content
Merck
All Photos(1)

Key Documents

B8299

Sigma-Aldrich

N-Butyldeoxynojirimycin

film (dried in situ), ≥98% (TLC)

Synonym(s):

Miglustat, NB-DNJ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21NO4
CAS Number:
Molecular Weight:
219.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Product Name

N-Butyldeoxynojirimycin, film (dried in situ)

Assay

≥98% (TLC)

Quality Level

form

film (dried in situ)

solubility

water: 9.80-10.20 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

CCCCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO

InChI

1S/C10H21NO4/c1-2-3-4-11-5-8(13)10(15)9(14)7(11)6-12/h7-10,12-15H,2-6H2,1H3/t7-,8+,9-,10-/m1/s1

InChI key

UQRORFVVSGFNRO-UTINFBMNSA-N

Gene Information

human ... UGCG(7357)

General description

N-Butyldeoxynojirimycin is an alkylated product of imino sugar deoxynojirimycin.

Application

N-Butyldeoxynojirimycin has been used:
  • in the inhibition of glycolipid synthesis in neuroblastoma cells
  • in the inhibition the ceramide-specific glycosyltransferase in hepatocytes
  • in the inhibition of β-glucosidase (GBA2) using fluorescence- activity assay in human embryonic kidney (HEK293) cells.

Biochem/physiol Actions

α-glucosidase Inhibitor
N-Butyldeoxynojirimycin is an inhibitor of glucosyltransferase and α-glucosidases. N-Butyldeoxynojirimycin, also known as misglustat, reduces glycolipid levels by substrate reduction therapy (SRT) and is effectively used for the treatment of glycosphingolipid lysosomal storage disorder, Gaucher disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

How adhesion/growth-regulatory galectins-1 and-3 attain cell specificity: case study defining their target on neuroblastoma cells (SK-N-MC) and marked affinity regulation by affecting microdomain organization of the membrane
Kopitz J, et al.
IUBMB Life, 62(8), 624-628 (2010)
Imiglucerase in the treatment of Gaucher disease: a history and perspective
Deegan PB and Cox TM
Drug design, development and therapy, 6, 81-81 (2012)
Andrés Felipe Leal et al.
International journal of molecular sciences, 21(17) (2020-09-02)
GM2 gangliosidoses are a group of pathologies characterized by GM2 ganglioside accumulation into the lysosome due to mutations on the genes encoding for the β-hexosaminidases subunits or the GM2 activator protein. Three GM2 gangliosidoses have been described: Tay-Sachs disease, Sandhoff
Beom Hee Lee et al.
Medicine, 96(45), e8492-e8492 (2017-11-16)
Gaucher disease (GD) is caused by a deficiency in the lysosomal enzyme glucocerebrosidase. Enzyme replacement therapy (ERT) is recommended for clinical improvement. The efficacy and safety of a new imiglucerase, Abcertin, were assessed in 7 Egyptian patients with treatment-naïve type
The role of the iminosugar N-butyldeoxynojirimycin (miglustat) in the management of type I (non-neuronopathic) Gaucher disease: a position statement
Cox TM, et al.
Journal of inherited metabolic disease, 26(6), 513-526 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service