Merck
Skip To
B7907
Nalgene® Lab Quality narrow-mouth HDPE bottles
high-density polyethylene, capacity 1,000 mL, with cap, translucent
Synonym(s):
HDPE bottles, nalgene bottle, nalgene bottles, Nalgene 2002-0032, laboratory container, vessel
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
About This Item
NACRES:
NB.17
UNSPSC Code:
41121800
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
high-density polyethylene , polypropylene cap, polypropylene screw closure, round bottle, translucent
joint
Joints threaded neck (38)
description
Not autoclavable
feature
with cap
packaging
pack of 6 ea
manufacturer/tradename
Nalgene® 2002-0032
capacity
1,000 mL
General description
Store, ship, and package liquids using highly reliable and durable Nalgene® narrow-mouth HDPE Lab Quality bottles with closure. These semi-rigid, translucent, all-purpose bottles are specifically designed for long-term, demanding lab use and offer excellent chemical resistance. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features and Benefits
  • Nalgene® narrow-mouth lab-quality bottles are ideal for storing and transporting liquids or dry chemicals in the lab.
  • Highly resistant to chemicals, these bottles are excellent for long-term use.
  • Includes linerless polypropylene leak-proof closures for preventing sample loss.
  • Uniform wall thickness protects against splitting or puncturing.
  • Bottles can withstand temperatures in a freezer up to -100°C.
  • Manufactured from non-toxic materials.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
B7532B3533B7782
material
high-density polyethylene , polypropylene screw closure, translucent , polypropylene cap, round bottle
material
high-density polyethylene , polypropylene cap, polypropylene screw closure, round bottle, translucent
material
high-density polyethylene , polypropylene cap, polypropylene screw closure, round bottle, translucent
material
high-density polyethylene , polypropylene cap, polypropylene screw closure, round bottle, translucent
manufacturer/tradename
Nalgene® 2002-0032
manufacturer/tradename
Nalgene® 2002-0002
manufacturer/tradename
Nalgene® 2002-0016
manufacturer/tradename
Nalgene® 2002-0008
feature
with cap
feature
with cap
feature
with cap
feature
with cap
capacity
1,000 mL
capacity
60 mL
capacity
500 mL
capacity
250 mL
joint
Joints threaded neck (38)
joint
Joints threaded neck (20)
joint
Joints threaded neck (28)
joint
Joints threaded neck (24)
packaging
pack of 6 ea
packaging
pack of 12 ea
packaging
pack of 12 ea
packaging
pack of 12 ea
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)