Skip to Content
Merck
All Photos(1)

Key Documents

B7276

Sigma-Aldrich

Anti-Bovine Albumin antibody produced in rabbit

fractionated antiserum, lyophilized powder

Synonym(s):

Anti-Albumin, bovine serum (BSA)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

bovine

packaging

vial of 2 mL lyophilized antiserum

technique(s)

indirect ELISA: 1:150,000

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

bovine ... Alb(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Serum albumin is the protein component of blood which regulates the colloid osmotic blood pressure and blood pH. Anti-bovine albumin antibody is used in study of in vivo hydrolysis of various bovine and human whey proteins which are measured by electroimmunoassay. Rabbit anti–bovine serum albumin antibodies react specifically with bovine serum albumin.

Immunogen

Purified BSA.

Application

Anti-bovine albumin antibody can be used in immunoaffinity purification and Ouchterlony double diffusion.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 mL deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Lindberg et al.
Journal of pediatric gastroenterology and nutrition, 24(5), 537-543 (1997-05-01)
There is limited information in the literature on the capacity of the preterm infant to digest human and bovine milk proteins. We therefore studied in vivo the luminal phase of the hydrolysis of proteins in human milk, human milk fortifier
E Shacter et al.
Blood, 82(9), 2853-2864 (1993-11-01)
Interleukin-6 (IL-6) is a multifunctional cytokine that is elevated in vivo during acute infection, chronic inflammation, and some hematopoietic malignancies. To understand how IL-6 becomes elevated in vivo, it is important to identify factors that can stimulate its secretion from
Xuanhong Cheng et al.
Lab on a chip, 10(8), 1079-1085 (2010-04-02)
Protein micropatterns have applications in fundamental life sciences and clinical medicine. In this work, we present a new technique to create 2-D protein micropatterns by local activation of a thin film of thermoresponsive plasma-deposited poly(N-isopropylacrylamide) (ppNIPAM) using a computer-controlled infrared
Carina Hillgruber et al.
The Journal of investigative dermatology, 134(1), 77-86 (2013-07-03)
Von Willebrand factor (VWF), a key player in hemostasis, is increasingly recognized as a proinflammatory protein. Here, we found a massive accumulation of VWF in skin biopsies of patients suffering from immune complex (IC)-mediated vasculitis (ICV). To clarify the impact
Lipoproteins, Apolipoproteins, and Lipases.
Verne N. Schumaker
Adv. Prot. Chem., 45, 1-459 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service