Skip to Content
Merck
All Photos(1)

Key Documents

B6891

Sigma-Aldrich

BME Vitamins 100x solution

BioReagent, sterile-filtered, suitable for cell culture

Synonym(s):

Vitamins mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352205
NACRES:
NA.75

sterility

sterile-filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

color

yellow to dark yellow

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

BME Vitamins 100x solution has been used as a component of M9 minimal medium for 15N-labeled protein expression. It has also been used as a supplement in neurobasal-A medium for organotypic hippocampal cultures.
BME Vitamins 100x solution is used in MOPS modified medium (MMM). It has been used as a supplement in several growth media.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca J Edgar et al.
Nature chemical biology, 15(5), 463-471 (2019-04-03)
Cell wall glycopolymers on the surface of Gram-positive bacteria are fundamental to bacterial physiology and infection biology. Here we identify gacH, a gene in the Streptococcus pyogenes group A carbohydrate (GAC) biosynthetic cluster, in two independent transposon library screens for
Alex P Lundin et al.
Journal of bacteriology, 202(9) (2020-02-20)
Bacterial microcompartments (MCPs) are widespread protein-based organelles composed of metabolic enzymes encapsulated within a protein shell. The function of MCPs is to optimize metabolic pathways by confining toxic and/or volatile pathway intermediates. A major class of MCPs known as glycyl
Micah Steffek et al.
Biochemistry (2022-01-06)
Autophagy-related proteins (Atgs) drive the lysosome-mediated degradation pathway, autophagy, to enable the clearance of dysfunctional cellular components and maintain homeostasis. In humans, this process is driven by the mammalian Atg8 (mAtg8) family of proteins comprising the LC3 and GABARAP subfamilies.
Yang Liu et al.
Journal of virology, 90(21), 9983-9996 (2016-08-26)
The P[19] genotype belongs to the P[II] genogroup of group A rotaviruses (RVs). However, unlike the other P[II] RVs, which mainly infect humans, P[19] RVs commonly infect animals (pigs), making P[19] unique for the study of RV diversity and host
Katharina Ludt et al.
Journal of bacteriology, 200(17) (2018-06-27)
Replication initiation in archaea involves a protein named ORC, Cdc6, or ORC1/Cdc6, which is homologous to the eukaryotic origin recognition complex (ORC) proteins and to the eukaryotic Cdc6. Archaeal replication origins are comprised of origin repeat regions and adjacent orc

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service