Skip to Content
Merck
All Photos(1)

Key Documents

B5556

Sigma-Aldrich

Bay 11-7082

≥98% (HPLC), powder, NF-kB/TNF-α inhibitor

Synonym(s):

(E)-3-(4-Methylphenylsulfonyl)-2-propenenitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2S
CAS Number:
Molecular Weight:
207.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Bay 11-7082, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: >20 mg/mL
H2O: insoluble

SMILES string

Cc1ccc(cc1)S(=O)(=O)\C=C\C#N

InChI

1S/C10H9NO2S/c1-9-3-5-10(6-4-9)14(12,13)8-2-7-11/h2-6,8H,1H3/b8-2+

InChI key

DOEWDSDBFRHVAP-KRXBUXKQSA-N

Application

Bay 11-7082 has been used as:
  • a nuclear factor-kappa B (NF-kB) inhibitor to verify the action of the NF-kB signaling pathway in the production of interleukin (IL)-8[1]
  • a nuclear factor-kappa B (NF-kB) inhibitor to study the role of NF-kB activation in Mycoplasma hyorhinis -induced epithelial-mesenchymal transition (EMT) and cell migration[2]
  • a nod-like receptor family pyrin domain containing 3 (NLRP3) selective inhibitor to examine its effects on liver inflammation in mice after hematopoietic stem cell transplantation[3]

Biochem/physiol Actions

Bay 11-7082 acts as a selective inhibitor for nod-like receptor family pyrin domain containing 3 (NLRP3) inflammasome pathway.[4] In addition, to the inhibition of nuclear factor-kappa B (NF-kB), Bay 11-7082 also triggers apoptosis in anucleated erythrocytes[5], human T-cell leukemia virus type I (HTLV-I)-infected T-cell lines and primary adult T-cell leukemia cells.[6]
Bay 11-7082 is an inhibitor of cytokine-induced IκB-α phosphorylation.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoping Cheng et al.
OncoTargets and therapy, 9, 7563-7571 (2016-12-23)
α-Catenin is an important molecule involved in the maintenance of cell-cell adhesion and a prognostic marker in cancer since its expression is essential for preventing cancer metastasis. However, the mechanism that leads to the downregulation of α-catenin in cancer progression
P Luz-Crawford et al.
Annals of the rheumatic diseases, 75(12), 2166-2174 (2016-03-11)
To define how peroxisome proliferator-activated receptor (PPAR) β/δ expression level in mesenchymal stem cells (MSCs) could predict and direct both their immunosuppressive and therapeutic properties. PPARβ/δ interacts with factors such as nuclear factor-kappa B (NF-κB) and regulates the expression of
Busulfan and cyclosphamide induce liver inflammation through NLRP3 activation in mice after hematopoietic stem cell transplantation
Qiao J, et al.
Scientific reports, 5, 17828-17828 (2015)
Atul Kumar et al.
Scientific reports, 7(1), 9535-9535 (2017-08-27)
Chronic myeloid leukemia (CML) is characterized by abnormal proliferation of myeloid cells which when untreated leads to bone marrow failure. Imatinib mesylate (IM) is the first line of therapy for treatment of CML and results in remission in most cases.
Adam G Peres et al.
Journal of leukocyte biology, 102(6), 1471-1480 (2017-10-12)
The aryl hydrocarbon receptor (AHR) is a ligand-activated transcription factor that triggers a broad response, which includes the regulation of proinflammatory cytokine production by monocytes and macrophages. AHR is negatively regulated by a set of genes that it transcriptionally activates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service