Skip to Content
Merck
All Photos(1)

Key Documents

B3563

Sigma-Aldrich

Bifonazole

≥98% (HPLC)

Synonym(s):

1-(p,α-Diphenylbenzyl)imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18N2
CAS Number:
Molecular Weight:
310.39
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

Bayer

storage temp.

room temp

SMILES string

c1ccc(cc1)C(c2ccc(cc2)-c3ccccc3)n4ccnc4

InChI

1S/C22H18N2/c1-3-7-18(8-4-1)19-11-13-21(14-12-19)22(24-16-15-23-17-24)20-9-5-2-6-10-20/h1-17,22H

InChI key

OCAPBUJLXMYKEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some Gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.
Bifonazole is used to treat several infections such as onychomycoses, otomycoses, erythrasma, psoriasis, sebopsoriasis, seborrhoeic dermatitis and rosacea. Bifonazole is also used as an important drug to treat melanoma.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clotrimazole and bifonazole detach hexokinase from mitochondria of melanoma cells
Penso J and Beitner R
European Journal of Pharmacology, 342(1), 113-117 (1998)
Sai Hu et al.
International journal of molecular medicine, 47(5) (2021-03-12)
Radioresistance is the predominant cause for radiotherapy failure and disease progression, resulting in increased breast cancer‑associated mortality. Using gene expression signature analysis of the Library of Integrated Network‑Based Cellular Signatures (LINCS) and Gene Expression Omnibus (GEO), the aim of the
Yonghong Zhao et al.
The Journal of biological chemistry, 281(9), 5973-5981 (2005-12-24)
To better understand ligand-induced structural transitions in cytochrome P450 2B4, protein-ligand interactions were investigated using a bulky inhibitor. Bifonazole, a broad spectrum antifungal agent, inhibits monooxygenase activity and induces a type II binding spectrum in 2B4dH(H226Y), a modified enzyme previously
Salomé El Hage et al.
Archiv der Pharmazie, 344(6), 402-410 (2011-03-25)
Two series of chlorinated benzhydryl imidazole and triazole derivatives were synthesized and tested in vitro against representative strains of potent pathogenic bacteria (Staphylococcus aureus CIP 4.83, Escherichia hirae CIP 5855, Pseudomonas aeruginosa CIP 82118, Escherichia coli CIP 53126) and fungi
Shinichi Watanabe et al.
Mycoses, 49(3), 236-241 (2006-05-10)
The aim of the study was to compare the efficacy and safety of luliconazole 1% cream and bifonazole 1% cream as applied in the treatment of tinea pedis (interdigital-type and plantar-type). A multi-clinic, randomised single-blind, parallel group study with 34

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service