Skip to Content
Merck
All Photos(1)

Key Documents

B2186

Sigma-Aldrich

BL-1249

≥98% (HPLC)

Synonym(s):

(5,6,7,8-Tetrahydro-naphthalen-1-yl)-[2-(1H-tetrazol-5-yl)-phenyl]-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N5
CAS Number:
Molecular Weight:
291.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

pink to brown

solubility

DMSO: ~17.5 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

C1CCc2c(C1)cccc2Nc3ccccc3-c4nnn[nH]4

InChI

1S/C17H17N5/c1-2-8-13-12(6-1)7-5-11-15(13)18-16-10-4-3-9-14(16)17-19-21-22-20-17/h3-5,7,9-11,18H,1-2,6,8H2,(H,19,20,21,22)

InChI key

YYNRZIFBTOUICE-UHFFFAOYSA-N

General description

BL-1249 is a non-steroid anti-inflammatory drug and an inhibitor of the cyclooxygenase (COX) enzyme.

Application

BL-1249 has been used:
  • as an activator of mechano-gated K2P channel in contracted mouse ileum and colon tissues
  • as a TWIK related potassium channel (TREK2) modulator and in U-2 OS osteosarcoma cell line
  • as a TREK1 and TREK2 activator in mice.

Biochem/physiol Actions

BL-1249 also activates TWIK related potassium channel 2 (TREK2). It interacts with K2P channel at the negatively charged activator site and modulates channel opening.
BL-1249 is a putative activator of potassium TREK-1 channel.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruolin Ma et al.
Frontiers in physiology, 9, 157-157 (2018-03-23)
Gastrointestinal (GI) motility disorders such as irritable bowel syndrome (IBS) can occur when coordinated smooth muscle contractility is disrupted. Potassium (K+) channels regulate GI smooth muscle tone and are key to GI tract relaxation, but their molecular and functional phenotypes
Ehab Al-Moubarak et al.
Scientific reports, 9(1), 12394-12394 (2019-08-29)
A better understanding of the gating of TREK two pore domain potassium (K2P) channels and their activation by compounds such as the negatively charged activator, flufenamic acid (FFA) is critical in the search for more potent and selective activators of
Ruolin Ma et al.
Frontiers in pharmacology, 11, 434-434 (2020-04-23)
TREK-1 channel activity is a critical regulator of neuronal, cardiac, and smooth muscle physiology and pathology. The antidepressant peptide, spadin, has been proposed to be a TREK-1-specific blocker. Here we sought to examine the mechanism of action underlying spadin inhibition
Paul D Wright et al.
Biochemical and biophysical research communications, 520(1), 35-40 (2019-10-01)
TREK2 (KCNK10, K2P10.1) is a two-pore domain potassium (K2P) channel and a potential target for the treatment of pain. Like the majority of the K2P superfamily, there is currently a lack of useful pharmacological tools to study TREK2. Here we
Svetlana Tertyshnikova et al.
The Journal of pharmacology and experimental therapeutics, 313(1), 250-259 (2004-12-21)
BL-1249 [(5,6,7,8-tetrahydro-naphthalen-1-yl)-[2-(1H-tetrazol-5-yl)-phenyl]-amine] produced a concentration-dependent membrane hyperpolarization of cultured human bladder myocytes, assessed as either a reduction in fluorescence of the voltage-sensitive dye bis-(1,2-dibutylbarbituric acid)trimethine oxonol (EC50 = 1.26 +/- 0.6 microM) or by direct electrophysiological measurement (EC50 = 1.49

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service