Skip to Content
Merck
All Photos(1)

Documents

B175

Sigma-Aldrich

BW 723C86

≥98% (HPLC)

Synonym(s):

α-methyl-5-(2-thienylmethoxy)-1H-Indole-3-ethanamine monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N2OS · HCl
CAS Number:
Molecular Weight:
322.85
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 1 mg/mL, clear

SMILES string

Cl.CC(N)Cc1c[nH]c2ccc(OCc3cccs3)cc12

InChI

1S/C16H18N2OS.ClH/c1-11(17)7-12-9-18-16-5-4-13(8-15(12)16)19-10-14-3-2-6-20-14;/h2-6,8-9,11,18H,7,10,17H2,1H3;1H

InChI key

PYJBJMIBANAOFJ-UHFFFAOYSA-N

Gene Information

human ... HTR2B(3357)

Biochem/physiol Actions

5-HT2B serotonin receptor agonist.

Caution

Hygroscopic; photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G A Kennett et al.
British journal of pharmacology, 117(7), 1443-1448 (1996-04-01)
1. BW 723C86 (3 and 10 mg kg-1, s.c. 30 min pretest), a 5-HT2B receptor agonist, increased total interaction, but not locomotion in a rat social interaction test, a profile consistent with anxiolysis. 2. The effect of BW 723C86 in
G A Kennett et al.
Neuropharmacology, 36(2), 233-239 (1997-02-01)
The 5-HT2B receptor agonist, BW 723C86 (10 and 20 mg/kg s.c.), increased the time spent in feeding behaviour of freely-fed rats in observation cages over 15 min. BW 723C86 (20 and 50 mg/kg s.c. 30 min pre-test) also modestly increased
Raphael R Perim et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 314(5), R709-R715 (2018-02-01)
Intermittent spinal serotonin receptor activation elicits phrenic motor facilitation (pMF), a form of spinal respiratory motor plasticity. Episodic activation of either serotonin type 2 (5-HT2) or type 7 (5-HT7) receptors elicits pMF, although they do so via distinct cellular mechanisms
Yasmine Chabbi-Achengli et al.
The American journal of pathology, 186(4), 927-937 (2016-03-13)
Rheumatoid arthritis is a chronic disease that results in a disabling and painful condition as it progresses to destruction of the articular cartilage and ankylosis of the joints. Although the cause of the disease is still unknown, evidence argues that
Marcus Niebert et al.
PloS one, 6(7), e21395-e21395 (2011-07-27)
Neurons of the respiratory network in the lower brainstem express a variety of serotonin receptors (5-HTRs) that act primarily through adenylyl cyclase. However, there is one receptor family including 5-HT(2A), 5-HT(2B), and 5-HT(2C) receptors that are directed towards protein kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service