Skip to Content
Merck

B0908

Nalgene® centrifuge bottles, Style 3122

nominal capacity 1000 mL

Synonym(s):

nalgene bottle, polycarbonate bottle, Centrifuge containers, Laboratory centrifuge bottles, Nalgene 3122-1000

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.11
UNSPSC Code:
41121703
Material:
polycarbonate , round bottle, screw top cap, translucent bottle, translucent polycarbonate , translucent polypropylene screw closure
Parameter:
7100 × g max. RCF
Feature:
RCF 7,100 × g, with cap
Packaging:
case of 16, pack of 4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polycarbonate , round bottle, screw top cap, translucent bottle, translucent polycarbonate , translucent polypropylene screw closure

joint

Joints threaded neck (52.6)

feature

RCF 7,100 × g, with cap

packaging

case of 16, pack of 4

manufacturer/tradename

Nalgene® 3122-1000

parameter

7100 × g max. RCF

bottle diam. × L

98.17 mm × 169.7 mm

nominal capacity

1000 mL

General description

“Nalgene” polycarbonate centrifuge bottles are ideal for conducting large-volume cell harvesting, pelleting, and protein purification. These bottles are molded from transparent polycarbonate, providing clarity for easy visual monitoring.

Features and Benefits

  • Ideal for low-speed centrifugation of biological and chemical samples.
  • Use polypropylene sealing cap assemblies for enhanced leakproof assurance, especially at maximum speeds or when handling hazardous materials.
  • Autoclavable.
  • Offers excellent mechanical strength properties..

Other Notes

Fill the bottles to at least 80% of their total capacity to ensure proper performance.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library