Skip to Content
Merck
All Photos(1)

Key Documents

B0293

Sigma-Aldrich

Nunc® MAX-100 Cryostore storage boxes

MAX-100 w/ 10 x 10 divider, for cryovial size 1.0-1.8mL, 24/cs

Synonym(s):

cryogenic tube storage box, cryogenic vial storage box, cryotube storage box, cryovial storage box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123304
NACRES:
NB.22
Pricing and availability is not currently available.

material

polycarbonate

feature

Holds 100 x 1-1.8 mL cryovials

packaging

case of 24
pack of 4

manufacturer/tradename

Nunc 374187

technique(s)

cryopreservation: suitable

size

132x132x52 mm

suitability

suitable for (cryogenic storage)

Looking for similar products? Visit Product Comparison Guide

General description

  • MAX-100 fits 1.0 and 1.8 mL tubes
  • Use 10 x 10 dividers for internally threaded tubes in gas phase of nitrogen
  • Use 9 x 9 dividers for externally threaded tubes or tubes fitted with CryoFlex

Legal Information

CryoFlex is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service