Skip to Content
Merck
All Photos(1)

Key Documents

AXYTE1004BR

Pipette tips for Eppendorf® and Socorex pipettors

racked, size 1000 μL, blue, pkg of 3x1152tips/cs

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1000 μL
packaging:
pkg of 3x1152tips/cs
manufacturer/tradename:
Axygen Inc TE-1004-B-R
Corning

manufacturer/tradename

Axygen Inc TE-1004-B-R
Corning

packaging

pkg of 3x1152tips/cs

size

1000 μL

volume

1000 μL

color

blue

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Browse all pipette tips

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service